Merck
  • Identification and validation of dysregulated MAPK7 (ERK5) as a novel oncogenic target in squamous cell lung and esophageal carcinoma.
Identification and validation of dysregulated MAPK7 (ERK5) as a novel oncogenic target in squamous cell lung and esophageal carcinoma.
BMC cancer (2015-06-05)
Paul R Gavine, Mei Wang, Dehua Yu, Eva Hu, Chunlei Huang, Jenny Xia, Xinying Su, Joan Fan, Tianwei Zhang, Qingqing Ye, Li Zheng, Guanshan Zhu, Ziliang Qian, Qingquan Luo, Ying Yong Hou, Qunsheng Ji
ABSTRACT
MAPK7/ERK5 (extracellular-signal-regulated kinase 5) functions within a canonical three-tiered MAPK (mitogen activated protein kinase) signaling cascade comprising MEK (MAPK/ERK kinase) 5, MEKK(MEK kinase) 2/3 and ERK5 itself. Despite being the least well studied of the MAPK-modules, evidence supports a role for MAPK7-signaling in the pathology of several cancer types. Fluorescence in situ hybridization (FISH) analysis identified MAPK7 gene amplification in 4% (3/74) of non-small cell lung cancers (NSCLC) (enriched to 6% (3/49) in squamous cell carcinoma) and 2% (2/95) of squamous esophageal cancers (sqEC). Immunohistochemical (IHC) analysis revealed a good correlation between MAPK7 gene amplification and protein expression. MAPK7 was validated as a proliferative oncogenic driver by performing in vitro siRNA knockdown of MAPK7 in tumor cell lines. Finally, a novel MEK5/MAPK7 co-transfected HEK293 cell line was developed and used for routine cell-based pharmacodynamic screening. Phosphorylation antibody microarray analysis also identified novel downstream pharmacodynamic (PD) biomarkers of MAPK7 kinase inhibition in tumor cells (pMEF2A and pMEF2D). Together, these data highlight a broader role for dysregulated MAPK7 in driving tumorigenesis within niche populations of highly prevalent tumor types, and describe current efforts in establishing a robust drug discovery screening cascade.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mapk7
Sigma-Aldrich
Thymidine 5′-triphosphate sodium salt, ≥96%
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPK7
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)