Merck
  • Surface treatment of titanium by a polydimethylsiloxane coating on bond strength of resin to titanium.
Surface treatment of titanium by a polydimethylsiloxane coating on bond strength of resin to titanium.
Journal of the mechanical behavior of biomedical materials (2014-12-03)
Christie Ying Kei Lung, Dan Liu, Jukka Pekka Matinlinna
ABSTRACT
To evaluate the in vitro effect of titanium surface treatment by a polydimethylsiloxane coating on the shear bond strength of a resin composite cement to titanium. The titanium samples (40×30mm) were divided into 4 groups (n=12). A control group was surface treated by sandblasting using 110µm silica-coated alumina powder at a constant pressure of 300kPa for 15s/1cm(2). For other three test groups, a polydimethylsiloxane silicone grease was applied onto the surface. The samples were subjected to thermal treatment in air at temperatures of 800°C, 1000°C, and 1100°C for 2h. A silane coupling agent was then applied and a resin composite stub was bonded using a polyethylene mold. The samples were submitted to three different storage conditions: dry storage, water storage in deionized water at 37°C for 30 days and thermo-cycled for 6000 cycles between 5.0 and 55.0°C. The shear bond strengths of all test groups were determined using a universal testing machine. Data were analyzed by two-way ANOVA and Tukey HSD (p<0.05). The surface morphology of titanium and elemental composition were examined with SEM and EDAX. The results showed that there was a significant difference for different surface treatments (p<0.001) and different storage conditions (p<0.01) on the mean shear bond strengths. EDAX analysis showed there was a change of elemental composition of titanium surface after thermal treatment of the coating. Surface treatment of titanium with a polydimethylsiloxane coating at 1000°C and 1100°C curing provides sufficient resin bonding for clinical services.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm