Merck
  • Estrogen-related receptor α is required for efficient human cytomegalovirus replication.
Estrogen-related receptor α is required for efficient human cytomegalovirus replication.
Proceedings of the National Academy of Sciences of the United States of America (2014-12-17)
Jesse Hwang, John G Purdy, Kai Wu, Joshua D Rabinowitz, Thomas Shenk
ABSTRACT
An shRNA-mediated screen of the 48 human nuclear receptor genes identified multiple candidates likely to influence the production of human cytomegalovirus in cultured human fibroblasts, including the estrogen-related receptor α (ERRα), an orphan nuclear receptor. The 50-kDa receptor and a 76-kDa variant were induced posttranscriptionally following infection. Genetic and pharmacological suppression of the receptor reduced viral RNA, protein, and DNA accumulation, as well as the yield of infectious progeny. In addition, RNAs encoding multiple metabolic enzymes, including enzymes sponsoring glycolysis (enolase 1, triosephosphate isomerase 1, and hexokinase 2), were reduced when the function of ERRα was inhibited in infected cells. Consistent with the effect on RNAs, a substantial number of metabolites, which are normally induced by infection, were either not increased or were increased to a reduced extent in the absence of normal ERRα activity. We conclude that ERRα is needed for the efficient production of cytomegalovirus progeny, and we propose that the nuclear receptor contributes importantly to the induction of a metabolic environment that supports optimal cytomegalovirus replication.
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
XCT790, ≥98% (HPLC), solid
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder