Merck
  • Effects of vitexin on the pharmacokinetics and mRNA expression of CYP isozymes in rats.
Effects of vitexin on the pharmacokinetics and mRNA expression of CYP isozymes in rats.
Phytotherapy research : PTR (2014-12-03)
Xin-shuai Wang, Xiao-chen Hu, Gui-ling Chen, Xiang Yuan, Rui-na Yang, Shuo Liang, Jing Ren, Jia-chun Sun, Guo-qiang Kong, She-gan Gao, Xiao-shan Feng
ABSTRACT
In traditional therapy with Chinese medicine, vitexin has several pharmacological properties, including antinociceptive, antispasmodic, antioxidant, antimyeloperoxidase, and α-glucosidase inhibitory activities. Recently, vitexin was shown to protect the heart against ischemia/reperfusion injury in an in vitro model by inhibiting apoptosis. The purpose of this study was to find out whether vitexin influences the effect on rat cytochrome P450 (CYP) enzymes (CYP1A2, CYP2C11, and CYP3A1) by using cocktail probe drugs in vivo; the influence on the levels of CYP mRNA was also studied. A cocktail solution at a dose of 5 mL/kg, which contained phenacetin (10 mg/kg), tolbutamide (1 mg/kg), and midazolam (5 mg/kg), was given as oral administration to rats treated with short or long period of intravenous vitexin via the caudal vein. Blood samples were collected at a series of time points, and the concentrations of probe drugs in plasma were determined by HPLC-mass spectrometry (MS)/MS. The corresponding pharmacokinetic parameters were calculated by the software of DAS 2.0. In addition, real-time reverse transcription-polymerase chain reaction was performed to determine the effects of vitexin on the mRNA expression of CYP1A2, CYP2C11, and CYP3A1 in rat liver. Treatment with short or long period of vitexin had no effects on rat CYP1A2. However, CYP3A1 enzyme activity was inhibited by vitexin in a concentration-dependent and time-dependent manner. Furthermore, CYP2C11 enzyme activity was induced after short period treatment but inhibited after long period of vitexin treatment. The mRNA expression results were in accordance with the pharmacokinetic results. In conclusion, vitexin can either inhibit or induce activities of CYP2C11 and CYP3A1. Therefore, caution is needed when vitexin is co-administered with some CYP2C11 or CYP3A1 substrates in clinic, which may result in treatment failure and herb-drug interactions.
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Pricing and availability is not currently available.
Supelco
Methanol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol
Pricing and availability is not currently available.
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, Absolute - Acetone free
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Pricing and availability is not currently available.
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Pricing and availability is not currently available.
Supelco
Acetonitrile, analytical standard
Pricing and availability is not currently available.
Supelco
Tolbutamide, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Pricing and availability is not currently available.
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Pricing and availability is not currently available.