Merck
  • Administered circulating microparticles derived from lung cancer patients markedly improved angiogenesis, blood flow and ischemic recovery in rat critical limb ischemia.
Administered circulating microparticles derived from lung cancer patients markedly improved angiogenesis, blood flow and ischemic recovery in rat critical limb ischemia.
Journal of translational medicine (2015-04-19)
Jiunn-Jye Sheu, Fan-Yen Lee, Christopher Glenn Wallace, Tzu-Hsien Tsai, Steve Leu, Yung-Lung Chen, Han-Tan Chai, Hung-I Lu, Cheuk-Kwan Sun, Hon-Kan Yip
ABSTRACT
We hypothesized that lung cancer patient's circulating microparticles (Lc-MPs) could promote angiogenesis, blood flow in ischemic zone and ischemic recovery in rat critical limb ischemia (CLI). To investigate the impact of MP therapy on reversing the setting of CLI, adult-male Sprague-Dawley rats (n=50) equally randomized into sham control (SC) (group 1), SC-Lc-MPs (1.0 x 10(7) particles) (group 2), CLI (group 3), CLI-Hs-MPs (MPs from healthy-subject) (group 4), and CLI-Lc-MPs (group 5) were sacrificed by post-CLI day-14. In vitro study showed that Lc-MPs enhanced VEGFR2 expression, angiogenesis, nitric-oxide production, and endothelial cell proliferation (all p<0.005). By days 7 and 14, Laser Doppler showed significantly higher ischemic/normal blood-flow ratio in groups 1 and 2 compared with group 3, and was significantly higher in group 4 and further elevated in group 5 (p<0.0001). Numbers of small vessels and endothelial markers (CD31(+) and vWF(+) cells) and protein expressions (eNOS, CD31) exhibited a pattern identical to Lasre Doppler among the five groups (all p<0.001). Pro-angiogenic factors (VEGF, CXCR4, SDF-1α, HGF) at cellular and protein levels showed a significant step-wise increase from groups 1 and 2 to groups 3, 4, and 5 (all p<0.001). Protein expressions of fibrotic (Smad3, TGF-β) and apoptotic (mitochondrial Bax, cleaved caspase 3, and PARP) biomarkers displayed an opposite pattern compared to that of Laser Doppler, whereas the protein expressions of anti-fibrotic (Smad1/5, BMP-2) and anti-apoptotic (Bcl-2) biomarkers showed an identical pattern compared with that of Laser Doppler among groups 1 to 3, and 5 (all p<0.001). Administration of Lc-MPs augmented angiogenesis and restored blood flow in a rat of CLI.
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, analytical standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%