Merck
  • Characterization of mesenchymal progenitor cell populations from non-epithelial oral mucosa.
Characterization of mesenchymal progenitor cell populations from non-epithelial oral mucosa.
Oral diseases (2014-09-03)
S Matsumura, K Higa, T Igarashi, S Takaichi, M Tonogi, N Shinozaki, J Shimazaki, G-y Yamane
ABSTRACT
The characteristics of cell populations extracted from oral mucosal non-epithelial tissues and their ability to differentiate were evaluated in vitro as a potential source of cells for mandibular and corneal regeneration. Oral mucosal non-epithelial cells (OMNECs) were extracted from tissue samples and were studied by flow cytometry and RT-PCR. Cells differentiating into osteoblasts, adipocytes, chondrocytes, neurocytes, or keratocytes were characterized by RT-PCR and cell staining. OMNECs expressed CD44, CD90, CD105, CD166, and STRO-1 antigens, which are markers for mesenchymal stem cells. In addition, Oct3/4, c-Myc, Nanog, KLF4, and Rex, which are expressed by embryonic or pluripotent stem cells, were detected by RT-PCR. Expression of CD49d, CD56, and PDGFRα, proteins closely associated with the neural crest, was observed in OMNECs, as was expression of Twist1, Sox9, Snail1 and Snail2, which are early neural crest and neural markers. Specific differentiation markers were expressed in OMNECs after differentiation into osteoblasts, adipocytes, chondrocytes, or keratocytes. Populations of OMNECs may contain both mesenchymal stem cells and neural crest origin cells and are a potential cell source for autologous regeneration of mandibular or corneal stroma.
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR