Merck
  • Stability of a liposomal formulation containing lipoyl or dihydrolipoyl acylglycerides.
Stability of a liposomal formulation containing lipoyl or dihydrolipoyl acylglycerides.
Journal of liposome research (2014-03-22)
Joseph A Laszlo, Kervin O Evans, David L Compton
ABSTRACT
The acylglycerides of lipoic and dihydrolipoic acids may serve as slow-release sources for cutaneous delivery of these antioxidants when formulated in a liposomal vehicle. Testing was conducted to determine the storage stability of lipoyl glycerides in phospholipid-based liposomes. Lipoyl glycerides prepared by transesterification of lipoic acid with high oleic sunflower oil were incorporated into unilamellar liposomes comprised of soy phosphatidylcholine (soyPC) or dioleoylphosphatidylcholine (DOPC). Lipoyl glycerides were stable in soyPC at 4 °C (90% remaining after five weeks) and decayed with a half-life (t(½)) of 14 d at 40 °C. In contrast, lipoyl glycerides embedded in DOPC were completely stable for four weeks at 40 °C. Dihydrolipoyl glycerides in soyPC converted to lipoyl glycerides at 4 °C (t(½) = 14 d) over four weeks, and much more rapidly so at 40 °C (t(½) = 1 d). A hydroperoxide accumulation analysis indicated that lipoyl glycerides and dihydrolipoyl glycerides were modified or degraded while suppressing autoxidation of the polyunsaturated fatty acids present in soyPC. Dynamic light scattering measurements found that liposomes containing lipoyl glycerides or dihydrolipoyl glycerides did not undergo significant size changes for at least 48 d, indicating that inclusion of the lipoic acid derivatives did not induce vesicle aggregation. Substitution of the soyPC with DOPC, which is not readily subject to autoxidation, provided a much more stable storage environment for lipoyl glycerides. These findings confirm the expectation that phospholipid liposomes need to be oxidatively stable vehicles for dermal delivery of lipoic acid derivatives.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Dioleoylglycerol, ≥99%, mixture of 1,3- and 1,2-isomers, liquid
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
USP
Alpha Lipoic Acid, United States Pharmacopeia (USP) Reference Standard
Thioctic acid containing impurity B, European Pharmacopoeia (EP) Reference Standard
Thioctic acid for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Thioctic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)