Merck
  • Biocompatible microemulsion modifies the tissue distribution of doxorubicin.
Biocompatible microemulsion modifies the tissue distribution of doxorubicin.
Journal of pharmaceutical sciences (2014-08-08)
Caroline Damico Candido, Michel Leandro Campos, Juliana Uruguay Correa Vidigal Assumpção, Kelly Chrystina Pestana, Elias Carvalho Padilha, Iracilda Zeppone Carlos, Rosângela Gonçalves Peccinini
ABSTRACT
The incorporation of doxorubicin (DOX) in a microemulsion (DOX-ME) has shown beneficial consequences by reducing the cardiotoxic effects of DOX. The aim of this study was to determine the distribution of DOX-ME in Ehrlich solid tumor (EST) and the heart, and compare it with that of free DOX. The distribution study was conducted with female Swiss mice with EST (n = 7 per group; 20-25 g). Animals received a single dose (10 mg/kg, i.p.) of DOX or DOX-ME 7 days after tumor inoculation. Fifteen minutes after administration, the animals were sacrificed, and the tumor and heart tissues were taken for immediate analysis by ultra-performance liquid chromatography. No difference was observed in DOX concentration in tumor tissue between DOX and DOX-ME administration. However, the most remarkable result in this study was the statistically significant reduction in DOX concentration in heart tissue of animals given DOX-ME. Mean DOX concentration in heart tissue was 0.92 ± 0.54 ng mg(-1) for DOX-ME and 1.85 ± 0.34 ng mg(-1) for free DOX. In conclusion, DOX-ME provides a better tissue distribution profile, with a lower drug concentration in heart tissue but still comparable tumor drug concentration, which indicates that antitumor activity would not be compromised.
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Sodium oleate, ≥95% (capillary GC)
Sigma-Aldrich
Sodium oleate, ≥99%
Daunorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Daunorubicin hydrochloride, ≥90% (HPLC)
Sigma-Aldrich
Sodium oleate, ≥82% (fatty acids), powder
Sigma-Aldrich
Daunorubicin hydrochloride, meets USP testing specifications