Merck
  • Vortex-assisted low density solvent liquid-liquid microextraction and salt-induced demulsification coupled to high performance liquid chromatography for the determination of five organophosphorus pesticide residues in fruits.
Vortex-assisted low density solvent liquid-liquid microextraction and salt-induced demulsification coupled to high performance liquid chromatography for the determination of five organophosphorus pesticide residues in fruits.
Talanta (2014-12-06)
Ketsarin Seebunrueng, Yanawath Santaladchaiyakit, Supalax Srijaranai
ABSTRACT
A simple and rapid microextraction method, vortex-assisted low density solvent liquid-liquid microextraction and salt-induced demulsification (VLLME-SID) coupled to high performance liquid chromatography (HPLC) was developed for the determination of organophosphorus pesticide (OPP) residues in fruits. The studied OPPs were azinphos-methyl, parathion-methyl, fenitrothion, diazinon and chlorpyrifos. For VLLME-SID, a mixture of low density solvents (1-dodecanol and hexane) was used as the extraction solvent under vortex agitation for enhancing dispersion. After complete dispersion, the emulsion was formed and the OPPs were extracted into extraction solvent droplets. Then, the emulsion was quickly broken up into two clear phases after the addition of AlCl3 as a demulsifier. Centrifugation was not required in this procedure. Under the optimal conditions, high enrichment factors (180-282), low limit of detections (LODs) (0.05-1 ng mL(-1)) and good precision (RSD≤7% for retention time and peak area) were obtained. The proposed method was successfully applied to the analysis of OPP residues in fruit samples (watermelon, grape, and cantaloupe). The LODs for samples were in the range 0.0006-0.0015 mg kg(-1) which are below the established EU-MRLs (0.01-0.3 mg kg(-1)). Good recoveries were also obtained (80-104%).
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Acetic acid, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
1-Undecanol, 97%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Supelco
Fenitrothion, PESTANAL®, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard