Merck
  • Design and in vitro evaluation of biocompatible dexamethasone-loaded nanoparticle dispersions, obtained from nano-emulsions, for inhalatory therapy.
Design and in vitro evaluation of biocompatible dexamethasone-loaded nanoparticle dispersions, obtained from nano-emulsions, for inhalatory therapy.
Colloids and surfaces. B, Biointerfaces (2014-12-02)
Cristina Fornaguera, Meritxell Llinàs, Conxita Solans, Gabriela Calderó
ABSTRACT
Polymeric nanoparticle dispersions containing dexamethasone (DXM) have been prepared from O/W nano-emulsions of the water/polysorbate 80/[4 wt% poly(lactide-co-glycolide) acid+0.18 wt% DXM in ethyl acetate] system by a low-energy method at 25°C. Nano-emulsions were formed at O/S ratios between 45/55 and 72/25 and water contents above 70 wt% by the phase inversion composition (PIC) method. The mean hydrodynamic diameter of nano-emulsions with a constant water content of 90 wt% and O/S ratios from 50/50 to 70/30 was below 350 nm as assessed by dynamic light scattering. The nanoparticles obtained from these nano-emulsions (by solvent evaporation) showed mean diameters of around 130 nm, as determined by transmission electron microscopy image analysis. Therapeutic concentrations of DXM were encapsulated in the nano-emulsions prior to nanoparticle preparation. DXM entrapment efficiency of the nanoparticle dispersion (above 74 wt%) decreased at increasing O/S ratios of the precursor nano-emulsion while DXM loading, which was around 10 mg/100 mL, showed the reverse tendency. DXM release from nanoparticle dispersions was about an order of magnitude slower than from an aqueous solution. In vitro studies performed in a lung carcinoma cell line and in vitro haemolysis studies performed in red blood cells revealed a dose-dependent toxicity and haemolytic response, respectively. The as-prepared nanoparticle dispersions were non-toxic up to a concentration of 40 μg/mL and non-haemolytic up to a concentration of 1 mg/mL. After purification, nanoparticle dispersions were non-toxic up to a concentration of 90 μg/mL. These results allow concluding that these polymeric nanoparticle dispersions are good candidates for inhalatory therapy.
Product Description
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Supelco
Glycolic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%