Merck
  • Culture environment regulates amino acid turnover and glucose utilisation in human ES cells.
Culture environment regulates amino acid turnover and glucose utilisation in human ES cells.
Reproduction, fertility, and development (2013-06-14)
Joy Rathjen, Christine Yeo, Charlotte Yap, Boon Siang Nicholas Tan, Peter D Rathjen, David K Gardner
ABSTRACT
Human embryonic stem (ES) cells have been proposed as a renewable source of pluripotent cells that can be differentiated into various cell types for use in research, drug discovery and in the emerging area of regenerative medicine. Exploitation of this potential will require the development of ES cell culture conditions that promote pluripotency and a normal cell metabolism, and quality control parameters that measure these outcomes. There is, however, relatively little known about the metabolism of pluripotent cells or the impact of culture environment and differentiation on their metabolic pathways. The effect of two commonly used medium supplements and cell differentiation on metabolic indicators in human ES cells were examined. Medium modifications and differentiation were compared in a chemically defined and feeder-independent culture system. Adding serum increased glucose utilisation and altered amino acid turnover by the cells, as well as inducing a small proportion of the cells to differentiate. Cell differentiation could be mitigated by inhibiting p38 mitogen-activated protein kinase (p38 MAPK activity). The addition of Knockout Serum Replacer also increased glucose uptake and changed amino acid turnover by the cells. These changes were distinct from those induced by serum and occurred in the absence of detectable differentiation. Induction of differentiation by bone morphogenetic protein 4 (BMP4), in contrast, did not alter metabolite turnover. Deviations from metabolite turnover by ES cells in fully defined medium demonstrated that culture environment can alter metabolite use. The challenge remains to understand the impact of metabolic changes on long-term cell maintenance and the functionality of derived cell populations.
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ≥99% (titration)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Imidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole, ≥99% (titration), free-flowing, Redi-Dri
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O