Merck
Evolving therapies for myocardial ischemia/reperfusion injury.
Journal of the American College of Cardiology (2015-04-11)
Borja Ibáñez, Gerd Heusch, Michel Ovize, Frans Van de Werf
ABSTRACT
The damage inflicted on the myocardium during acute myocardial infarction is the result of 2 processes: ischemia and subsequent reperfusion (ischemia/reperfusion injury). During the last 3 decades, therapies to reduce ischemic injury (mainly reperfusion strategies) have been widely incorporated into clinical practice. The remarkable reduction in death rates achieved with these therapies has resulted in a shift in emphasis from efforts to reduce mortality to a focus on tackling the downstream consequence of survival: post-infarction heart failure. Infarct size is the main determinant of long-term mortality and chronic heart failure, and thus, the possibility of limiting the extent of necrosis during an ST-segment elevation myocardial infarction is of great individual and socioeconomic value. After the great success of therapies to reduce ischemic injury, the time has come to focus efforts on therapies to reduce reperfusion injury, but in the recent few years, few interventions have successfully passed the proof-of-concept stage. In this review, we examine the past, present, and future therapies to reduce ischemia/reperfusion injury.
Product Description
Supelco
Nitrite ion standard solution, for ion-selective electrodes, 0.1 M NO2-
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Supelco
Sodium nitrite, analytical standard
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard