Merck
  • Metal phthalocyanine: fullerene composite nanotubes via templating method for enhanced properties.
Metal phthalocyanine: fullerene composite nanotubes via templating method for enhanced properties.
Nanoscale research letters (2015-04-09)
Abdullah Haaziq Ahmad Makinudin, Muhamad Saipul Fakir, Azzuliani Supangat
ABSTRACT
The use of templating method to synthesize the vanadyl 2,9,16,23-tetraphenoxy-29H,31H-phthalocyanine (VOPcPhO):[6,6]-phenyl C71 butyric acid methyl ester (PC71BM) composite nanotubes is presented here. VOPcPhO is a p-type material and PC71BM is an n-type material which acts as an electron donor and electron acceptor, respectively. Both materials have been studied due to their potential applications as solar energy converter and organic electronics. High-resolution transmission electron microscope (HRTEM) and field emission scanning electron microscope (FESEM) images have shown the replication of the porous template diameter of approximately 200 nm with a superior incorporation of both VOPcPhO and PC71BM. VOPcPhO:PC71BM composite nanotubes showed the significant properties improvement if compared over their bulk heterojunction counterpart. UV-vis spectra of composite nanotubes show a shift to a longer wavelength at the absorption peaks. Significant quenching has been attained by the photoluminescence spectra of VOPcPhO:PC71BM composite nanotubes which supports the redshift of UV-vis absorption spectra. Presumably, the photo-induced charge transfer and charge carrier dissociation can be enhanced from the VOPcPhO:PC71BM composite nanotubes rather than the bulk heterojunction.
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh