Merck
  • Protective effects of escin against indomethacin-induced gastric ulcer in mice.
Protective effects of escin against indomethacin-induced gastric ulcer in mice.
Toxicology mechanisms and methods (2014-08-20)
Tian Wang, Shanshan Zhao, Yucun Wang, Yujiao Yang, Le Yao, Liuxiang Chu, Hanhan Du, Fenghua Fu
ABSTRACT
Escin, a natural mixture of triterpenoid saponin isolated from the seed of the horse chestnut, is reported to have a potent antiulcer activity against ethanol-induced gastric mucosal lesions. This study investigated the possible mechanisms underlying the gastroprotective effect of escin against indomethacin-induced gastric ulcer in mice. Gastric ulceration was induced by a single intragastric administration of indomethacin (18 mg/kg). The mice underwent intragastric treatment with escin at doses of 0.45, 0.9 or 1.8 mg/kg. Gastric lesion was estimated morphometrically and histopathologically 6 h after the indomethacin administration. The antioxidative parameters in gastric mucosa were measured. Moreover, the activity of myeloperoxidase and the contents of TNF-α, P-selectin and VCAM-1 in gastric tissues were determined. The results showed that escin protected gastric tissues against indomethacin-induced gastropathy as demonstrated from a reduction in the ulcer index and an attenuation of histopathologic changes. Escin caused significant reductions of the contents of malondialdehyde, TNF-α, P-selectin, VCAM-1 and myeloperoxidase activity. The altered activities of superoxide dismutase, catalase and glutathione peroxidase in the stomach tissues were also ameliorated by escin treatment. The present study demonstrated that escin had a protective effect against indomethacin-induced gastric ulcer in mice, not only by virtue of its antioxidant potential, but also due to its anti-inflammatory effect.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Ammonium molybdate, meets USP testing specifications