Merck
  • Engineering the synthesis of silica-gold nano-urchin particles using continuous synthesis.
Engineering the synthesis of silica-gold nano-urchin particles using continuous synthesis.
Nanoscale (2014-09-27)
Víctor Sebastian, Seung-Kon Lee, Klavs F Jensen
ABSTRACT
Compared to freestanding nanoparticles, supported nanostructures typically show better mechanical stability as well as ease of handling. Unique shapes such as core-shells, raspberries and crescents have been developed on supported materials to gain improved chemical and optical properties along with versatility and tunability. We report the formation of hyper-branched gold structures on silica particles, silica-gold nano-urchin (SGNU) particles. Kinetic control of crystallization, fast mass transfer as well as a bumped surface morphology of the silica particles are important factors for the growth of gold branches on the silica support. Using a microfluidic platform, continuous synthesis of SGNUs is achieved with increased reaction rate (less than 12 min of residence time), better controllability and reproducibility than that obtained in batch synthesis. The hyper-branched gold structures display surface-enhanced Raman scattering (SERS).
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Supelco
L-Ascorbic acid, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Octane, anhydrous, ≥99%
Supelco
Hexadecane, analytical standard
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Supelco
Cyclohexane, analytical standard
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Supelco
Octane, analytical standard
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%