Merck
  • Cortisol level measurements in fingernails as a retrospective index of hormone production.
Cortisol level measurements in fingernails as a retrospective index of hormone production.
Psychoneuroendocrinology (2015-02-11)
Shuhei Izawa, Keiichi Miki, Masao Tsuchiya, Takeshi Mitani, Toru Midorikawa, Tatsuya Fuchu, Taiki Komatsu, Fumiharu Togo
ABSTRACT
The cortisol level in fingernails may reflect the hormone's cumulative production over a long period, but the notions have not been fully established. In this study, we investigated the association of cortisol in fingernails with cortisol accumulation over a long period (hair cortisol) and over a relatively short period (salivary cortisol). In study 1, hair and fingernail samples were collected from 58 middle-aged and elderly men. The cortisol level in hair samples was moderately associated with the level in fingernail samples (r = 0.29, p < 0.05 and rs = 0.36, p < 0.01). In study 2, 37 workers provided 4 saliva samples over the course of one day (at awakening, 30 min after awakening, before lunch, and after work) and another set a month later. Further, the workers were asked to provide fingernail samples during a six-month period. We found that the cortisol level in saliva over the whole day (area under the curve for cortisol) was moderately associated with the cortisol level measured in fingernail samples that were collected 4 months (r = 0.43, p < 0.05 and rs = 0.50, p < 0.01) and 5 months later (r = 0.45, p < 0.05 and rs = 0.53, p < 0.01). These results indicated that the cortisol level in fingernail samples might retrospectively represent hormone production during a given period. The cortisol level in fingernail samples may be useful in the investigation of the link between psychosocial stress and health.
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O