Merck
  • Structural characterization of lignin: a potential source of antioxidants guaiacol and 4-vinylguaiacol.
Structural characterization of lignin: a potential source of antioxidants guaiacol and 4-vinylguaiacol.
International journal of biological macromolecules (2015-01-21)
Mohammadali Azadfar, Allan Haiming Gao, Mahesh V Bule, Shulin Chen
ABSTRACT
The structure of lignin obtained from the ozone and soaking aqueous ammonia pretreatment of wheat straw has been characterized utilizing chemical analytical methods in order to reveal its antioxidant characteristics, including attenuated total reflectance-Fourier transform infrared spectroscopy (ATR-FTIR), pyrolysis-gas chromatography/mass spectrometry (Py-GC/MS), pyrolysis/tetramethylammonium hydroxide-gas chromatography/mass spectrometry (Py/TMAH-GC/MS), gel permeation chromatography (GPC), ultra violet-visible spectroscopy (UV-vis), and 1,1-diphenyl-2-picrylhydrazyl (DPPH) antioxidant evaluation assay. The results demonstrated that the isolated lignin is a ρ-hydroxyphenyl- guaiacyl-syringyl (H-G-S) lignin, with S/G ratio of 0.35 and significant amounts of phenol 2-methoxy (guaiacol) and phenol 2-methoxy-4-vinyl (4-vinylguaiacol). The Py-GC/MS and Py/TMAH-GC/MS pyrograms indicated that the major units in this lignin are derived from hydroxycinnamic acids. The GPC results revealed the molecular weight of the lignin was considerably low and also the FTIR analysis showed that the lignin possessed hydroxyl and methoxy functional groups; the factors led to the extracted lignin having a comparable antioxidant activity to that of currently used commercial antioxidants. The UV-vis and DPPH antioxidant assay results suggested a percentage of inhibition of the DPPH radicals in the following order: guaiacol (103.6 ± 1.36)>butylated hydroxytoluene (103.3 ± 1)>ferulic acid (102.6 ± 0.79)>pretreated lignin (86.9 ± 0.34).
Product Description
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetic acid, analytical standard
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Guaiacol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methoxyphenol, 97%
Sigma-Aldrich
trans-Ferulic acid, ≥99%