Skip to Content
Merck
All Photos(1)

Documents

Y0000619

Guaiacol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Methoxyphenol, Catechol monomethyl ether, Pyrocatechol monomethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)C6H4OH
CAS Number:
Molecular Weight:
124.14
Beilstein:
508112
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

vapor density

4.27 (vs air)

vapor pressure

0.11 mmHg ( 25 °C)

API family

guaiacol

manufacturer/tradename

EDQM

refractive index

n20/D 1.543 (lit.)

bp

205 °C (lit.)

mp

26-29 °C (lit.)

density

1.129 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccccc1O

InChI

1S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8H,1H3

InChI key

LHGVFZTZFXWLCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Guaiacol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Guaiacol, along with 2,4,6-trichloroanisole, is responsible for cork taint in wine. A method has been developed for extraction and quantitation of the two compounds.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Woo-Cheol Sim et al.
Biochemical pharmacology, 90(4), 414-424 (2014-06-24)
Collaborative regulation of liver X receptor (LXR) and sterol regulatory element binding protein (SREBP)-1 are main determinants in hepatic steatosis, as shown in both animal models and human patients. Recent studies indicate that selective intervention of overly functional LXRα in
Tao Liu et al.
The Analyst, 139(17), 4315-4321 (2014-07-06)
The rapid and sensitive detection of Alicyclobacillus acidoterrestris (AA) has become very important due to the frequent occurrence of fruit juice spoilage by AA. In the present study, using guaiacol, both as the metabolic product of AA related to its
Nathalie Allaman-Pillet et al.
Molecular cancer research : MCR, 13(1), 86-97 (2014-08-22)
Retinoblastoma is the most common pediatric intraocular neoplasm. While retinoblastoma development requires the inactivation of both alleles of the retinoblastoma tumor suppressor gene (RB1) in the developing retina, additional genomic changes are involved in tumor progression, which progressively lead to
Sonica Sondhi et al.
PloS one, 9(5), e96951-e96951 (2014-05-30)
A novel extracellular thermo-alkali-stable laccase from Bacillus tequilensis SN4 (SN4LAC) was purified to homogeneity. The laccase was a monomeric protein of molecular weight 32 KDa. UV-visible spectrum and peptide mass fingerprinting results showed that SN4LAC is a multicopper oxidase. Laccase
Guus van Rossum et al.
ChemSusChem, 7(1), 253-259 (2013-11-23)
The liquefaction of lignocellulosic biomass is studied for the production of liquid (transportation) fuels. The process concept uses a product recycle as a liquefaction medium and produces a bio-oil that can be co-processed in a conventional oil refinery. This all

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service