Merck
  • 3-Methyladenine can depress drug efflux transporters via blocking the PI3K-AKT-mTOR pathway thus sensitizing MDR cancer to chemotherapy.
3-Methyladenine can depress drug efflux transporters via blocking the PI3K-AKT-mTOR pathway thus sensitizing MDR cancer to chemotherapy.
Journal of drug targeting (2014-07-16)
Zhenyou Zou, Jing Zhang, Haiyang Zhang, Huan Liu, Zhiguo Li, Dan Cheng, Jun Chen, Lu Liu, Mengjie Ni, Yu Zhang, Jun Yao, Jun Zhou, Junjie Fu, Yong Liang
ABSTRACT
Multi-drug resistance (MDR) cancer is an intractable problem. Over-expression of drug efflux transporters such as ABCB1, ABCC1 and ABCG2 contributes to it, by which they pump drugs out of cells, and result in the decrease in the efficacy of chemotherapy. To reverse the cancer MDR, we used 3-methyladenine (3-MA) treatment on taxol or doxorubicin stressed MDR cell lines A2780DX5 and SGC7091R and xeno-tumor implanted mice. The results indicate that ABCB1, ABCC1 and ABCG2 were depressed, and the PI3K-AKT-mTOR pathway was blocked. Moreover, using FITC-labeled taxol as the indicator, we observed that the drug accumulation was enhanced in MDR cells and more cells were killed after 3-MA administration. Thus suggesting that 3-MA can reverse cancer MDR via depressing agent-efflux transporters.
Sigma-Aldrich
MISSION® esiRNA, targeting human PIK3R1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pik3r1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pik3ca
Sigma-Aldrich
MISSION® esiRNA, targeting human PIK3CA
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Rifampicin, European Pharmacopoeia (EP) Reference Standard
Paclitaxel natural for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals