Skip to Content
Merck
All Photos(1)

Key Documents

EHU039391

Sigma-Aldrich

MISSION® esiRNA

targeting human PIK3CA

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACCTTGTTCCAATCCCAGGTGGAATGAATGGCTGAATTATGATATATACATTCCTGATCTTCCTCGTGCTGCTCGACTTTGCCTTTCCATTTGCTCTGTTAAAGGCCGAAAGGGTGCTAAAGAGGAACACTGTCCATTGGCATGGGGAAATATAAACTTGTTTGATTACACAGACACTCTAGTATCTGGAAAAATGGCTTTGAATCTTTGGCCAGTACCTCATGGATTAGAAGATTTGCTGAACCCTATTGGTGTTACTGGATCAAATCCAAATAAAGAAACTCCATGCTTAGAGTTGGAGTTTGACTGGTTCAGCAGTGTGGTAAAGTTCCCAGATATGTCAGTGATTGAAGAGCATGCCAATTGGTCTGTATCCCGAGAAGCAGGATTTAGCTATTCCCACGCAGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiuling Liang et al.
Yonsei medical journal, 60(2), 182-190 (2019-01-23)
This study aimed to investigate the effects of PIK3CA on the sensitivity of acute B lymphocytic leukemia cells (Nalm-6 cells) to chemotherapy drugs. Children's normal B lymphocytes and Nalm-6 cells were cultured. Nalm-6 cells were transfected with PIK3CA siRNA (siPIK3CA
Zhao Wang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 43(6), 2505-2515 (2017-11-14)
Numerous studies have demonstrated that aberrant microRNA (miRNA) expression is involved in human disease including cancer. To date, the potential miRNAs regulating lung cancer growth and progression are not fully identified yet. In this study, the expression of miR-142-5p was
Shuke Ge et al.
Oncology research, 25(8), 1363-1371 (2017-03-02)
miR-152, as a tumor suppressor, has been reported to be downregulated in a number of cancer cell lines and tumor tissues, including breast cancer. This study aimed to investigate the role of miR-152 in human breast cancer and its underlying
Jun Xu et al.
Molecular medicine reports, 12(3), 4708-4712 (2015-06-24)
The expression of osteopontin (OPN) and vascular endothelial growth factor (VEGF) are associated with the severity of cartilage destruction in osteoarthritis. However, the biological connection between OPN and VEGF in osteoarthritis remains to be elucidated. The present study was performed
Xue Bai et al.
Oncology reports, 33(6), 3085-3092 (2015-05-13)
Cervical cancer is the second most common women carcinoma worldwide and the fourth leading cause of cancer-associated mortality in women. Butein, a bioactive flavonoid isolated from numerous native plants, has been shown to induce apoptosis and inhibits migration and invasion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service