Merck
  • Directed divergent evolution of a thermostable D-tagatose epimerase towards improved activity for two hexose substrates.
Directed divergent evolution of a thermostable D-tagatose epimerase towards improved activity for two hexose substrates.
Chembiochem : a European journal of chemical biology (2015-02-07)
Andreas Bosshart, Chee Seng Hee, Matthias Bechtold, Tilman Schirmer, Sven Panke
ABSTRACT
Functional promiscuity of enzymes can often be harnessed as the starting point for the directed evolution of novel biocatalysts. Here we describe the divergent morphing of an engineered thermostable variant (Var8) of a promiscuous D-tagatose epimerase (DTE) into two efficient catalysts for the C3 epimerization of D-fructose to D-psicose and of L-sorbose to L-tagatose. Iterative single-site randomization and screening of 48 residues in the first and second shells around the substrate-binding site of Var8 yielded the eight-site mutant IDF8 (ninefold improved kcat for the epimerization of D-fructose) and the six-site mutant ILS6 (14-fold improved epimerization of L-sorbose), compared to Var8. Structure analysis of IDF8 revealed a charged patch at the entrance of its active site; this presumably facilitates entry of the polar substrate. The improvement in catalytic activity of variant ILS6 is thought to relate to subtle changes in the hydration of the bound substrate. The structures can now be used to select additional sites for further directed evolution of the ketohexose epimerase.
Product Description
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, BioXtra, ≥99.0%
Sigma-Aldrich
D-(+)-Sorbose, ≥99%
Sigma-Aldrich
1,4-Dithioerythritol, BioReagent, ≥99.0%
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-Psicose, ≥95% (HPLC)
Sigma-Aldrich
L-(+)-Fructose, ≥97.0% (HPLC)
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0% (RT), BioUltra
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Fructose, European Pharmacopoeia (EP) Reference Standard
USP
Fructose, United States Pharmacopeia (USP) Reference Standard
USP
Galactitol, United States Pharmacopeia (USP) Reference Standard
Galactitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Dulcitol, ≥99% (GC)