Skip to Content
Merck
All Photos(2)

Key Documents

47748

Sigma-Aldrich

D-(−)-Fructose

tested according to Ph. Eur.

Synonym(s):

Laevulosum (Fructosum), D-Levulose, Fruit sugar

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$65.50

$65.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$65.50

About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1239004
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

$65.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

form

solid

impurities

≤0.5% water

color

colorless

useful pH range

5.0-7.0 (25 °C, 18 g/L)

mp

119-122 °C (dec.) (lit.)

solubility

alcohol: soluble 1 gm in 15 ml
methanol: soluble 1 gm in 14 ml
H2O: freely soluble
acetone: freely soluble (Hot)
acetone: slightly soluble
pyridine: soluble

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(−)-Fructose has been used as a test compound for investigating the enzyme that catalyzes the synthesis of activator for phosphofructokinase termed “activation factor” by utilizing ATP and D-fructose-6-P.[1]

Biochem/physiol Actions

D-(−)-Fructose can enhance mood and gastrointestinal disturbances in fructose malabsorbers.[2] It also possess metabolic and endocrine impact that shows that increased consumption of fructose is a contributing factor in the development of obesity and the accompanying metabolic abnormalities observed in the insulin resistance syndrome.[3]

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Furuya et al.
The Journal of biological chemistry, 256(14), 7109-7112 (1981-07-25)
We have recently discovered an activator for phosphofructokinase termed "activation factor" (Furuya, E., and Uyeda, K. (1980) Proc. Natl. Acad. Sci. U. S. A. 77, 5861-5864). In this paper, we investigated the enzyme responsible for its synthesis. We have found
Sharon S Elliott et al.
The American journal of clinical nutrition, 76(5), 911-922 (2002-10-26)
This review explores whether fructose consumption might be a contributing factor to the development of obesity and the accompanying metabolic abnormalities observed in the insulin resistance syndrome. The per capita disappearance data for fructose from the combined consumption of sucrose
M Ledochowski et al.
Scandinavian journal of gastroenterology, 35(10), 1048-1052 (2000-12-01)
Fructose malabsorption is characterized by the inability to absorb fructose efficiently. As a consequence fructose reaches the colon where it is broken down by bacteria to short fatty acids, CO2 and H2. Bloating, cramps, osmotic diarrhea and other symptoms of
Carlos A Roncal Jimenez et al.
Kidney international, 86(2), 294-302 (2013-12-18)
The epidemic of chronic kidney disease in Nicaragua (Mesoamerican nephropathy) has been linked with recurrent dehydration. Here we tested whether recurrent dehydration may cause renal injury by activation of the polyol pathway, resulting in the generation of endogenous fructose in
Jérémy Lothier et al.
Planta, 240(3), 629-643 (2014-07-16)
The first 6-fructan exohydrolase (6-FEH) cDNA from Lolium perenne was cloned and characterized. Following defoliation, Lp6 - FEHa transcript level unexpectedly decreased together with an increase in total FEH activity. Lolium perenne is a major forage grass species that accumulates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service