Merck
  • Effects of natural nuclear factor-kappa B inhibitors on anticancer drug efflux transporter human P-glycoprotein.
Effects of natural nuclear factor-kappa B inhibitors on anticancer drug efflux transporter human P-glycoprotein.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2015-03-18)
Tomohiro Nabekura, Takashi Hiroi, Tatsuya Kawasaki, Yuichi Uwai
ABSTRACT
Drug efflux transporter P-glycoprotein plays an important role in cancer chemotherapy. The nuclear factor-κB (NF-κB) transcription factors play critical roles in development and progression of cancer. In this study, the effects of natural compounds that can inhibit NF-κB activation on the function of P-glycoprotein were investigated using human MDR1 gene-transfected KB/MDR1 cells. The accumulation of daunorubicin or rhodamine 123, fluorescent substrates of P-glycoprotein, in KB/MDR1 cells increased in the presence of caffeic acid phenetyl ester (CAPE), licochalcone A, anacardic acid, celastrol, xanthohumol, magnolol, and honokiol in a concentration-dependent manner. In contrast, lupeol, zerumbone, thymoquinone, emodin, and anethol had no effects. The ATPase activities of P-glycoprotein were stimulated by CAPE, licochalcone A, anacardic acid, celastrol, xanthohumol, magnolol, and honokiol. Tumor necrosis factor (TNF)-α stimulated NF-κB activation was inhibited by CAPE, licochalcone A, anacardic acid, and xanthohumol. KB/MDR1 cells were sensitized to vinblastine cytotoxicity by CAPE, licochalcone A, anacardic acid, xanthohumol, magnolol, and honokiol, showing that these natural NF-κB inhibitors reverse multidrug resistance. These results suggest that natural compounds, such as CAPE, licochalcone A, and anacardic acid, have dual inhibitory effects on the anticancer drug efflux transporter P-glycoprotein and NF-κB activation, and may become useful to enhance the efficacy of cancer chemotherapy.
Sigma-Aldrich
Anacardic acid
Supelco
trans-Anethole, analytical standard
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
trans-Anethole, 99%
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
trans-Anethole, ≥99%, FCC, FG
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Supelco
Eugenol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Thymoquinone, ≥98%
Supelco
Thymoquinone, analytical standard
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
Eugenol, European Pharmacopoeia (EP) Reference Standard
Supelco
Emodin, analytical standard
Sigma-Aldrich
Licochalcone A, ≥96.0% (HPLC)
Supelco
Lupeol, analytical standard
Sigma-Aldrich
Maslinic acid, ≥98% (HPLC)
Sigma-Aldrich
Honokiol, ≥98% (HPLC), powder
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Emodin, from Frangula bark, ≥90% (HPLC)
Sigma-Aldrich
Lupeol, ≥94%
Sigma-Aldrich
Caffeic acid phenethyl ester, ≥97% (HPLC), powder
Supelco
Honokiol, analytical standard
Sigma-Aldrich
Rhodamine 123, BioReagent, for fluorescence, ≥85% (HPLC)
Honokiol, European Pharmacopoeia (EP) Reference Standard