Merck
  • Formulation pre-screening of inhalation powders using computational atom-atom systematic search method.
Formulation pre-screening of inhalation powders using computational atom-atom systematic search method.
Molecular pharmaceutics (2014-11-08)
Vasuki Ramachandran, Darragh Murnane, Robert B Hammond, Jonathan Pickering, Kevin J Roberts, Majeed Soufian, Ben Forbes, Sara Jaffari, Gary P Martin, Elizabeth Collins, Klimentina Pencheva
ABSTRACT
The synthonic modeling approach provides a molecule-centered understanding of the surface properties of crystals. It has been applied extensively to understand crystallization processes. This study aimed to investigate the functional relevance of synthonic modeling to the formulation of inhalation powders by assessing cohesivity of three active pharmaceutical ingredients (APIs, fluticasone propionate (FP), budesonide (Bud), and salbutamol base (SB)) and the commonly used excipient, α-lactose monohydrate (LMH). It is found that FP (-11.5 kcal/mol) has a higher cohesive strength than Bud (-9.9 kcal/mol) or SB (-7.8 kcal/mol). The prediction correlated directly to cohesive strength measurements using laser diffraction, where the airflow pressure required for complete dispersion (CPP) was 3.5, 2.0, and 1.0 bar for FP, Bud, and SB, respectively. The highest cohesive strength was predicted for LMH (-15.9 kcal/mol), which did not correlate with the CPP value of 2.0 bar (i.e., ranking lower than FP). High FP-LMH adhesive forces (-11.7 kcal/mol) were predicted. However, aerosolization studies revealed that the FP-LMH blends consisted of agglomerated FP particles with a large median diameter (∼4-5 μm) that were not disrupted by LMH. Modeling of the crystal and surface chemistry of LMH identified high electrostatic and H-bond components of its cohesive energy due to the presence of water and hydroxyl groups in lactose, unlike the APIs. A direct comparison of the predicted and measured cohesive balance of LMH with APIs will require a more in-depth understanding of highly hydrogen-bonded systems with respect to the synthonic engineering modeling tool, as well as the influence of agglomerate structure on surface-surface contact geometry. Overall, this research has demonstrated the possible application and relevance of synthonic engineering tools for rapid pre-screening in drug formulation and design.
Sigma-Aldrich
Budesonide, ≥99%
Sigma-Aldrich
D-Lactose monohydrate, tested according to Ph. Eur.
Sigma-Aldrich
D-Lactose monohydrate, ≥99% (HPLC), BioUltra
Sigma-Aldrich
D-Lactose monohydrate, ACS reagent
Sigma-Aldrich
D-Lactose monohydrate, ≥98.0% (HPLC)
Supelco
Budesonide, Pharmaceutical Secondary Standard; Certified Reference Material
Budesonide, European Pharmacopoeia (EP) Reference Standard
Supelco
Lactose (Monohydrate), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluticasone propionate, ≥98% (HPLC), powder
Supelco
Salbutamol, VETRANAL®, analytical standard
Sigma-Aldrich
Salbutamol
Fluticasone propionate, European Pharmacopoeia (EP) Reference Standard
USP
Lactose monohydrate, United States Pharmacopeia (USP) Reference Standard
USP
Fluticasone propionate, United States Pharmacopeia (USP) Reference Standard