Skip to Content
Merck
All Photos(1)

Key Documents

PHR1024

Supelco

Lactose (Monohydrate)

Lactose (Monohydrate)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

D-Lactose monohydrate, β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, Milk sugar

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$147.00

$147.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 09, 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$147.00

About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Beilstein:
3768231
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$147.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 09, 2025Details


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001750
traceable to USP 1356701

API family

lactose

CofA

current certificate can be downloaded

analyte chemical class(es)

oligosaccharides

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
detection
environmental
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

O.OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1

InChI key

HBDJFVFTHLOSDW-XBLONOLSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Lactose (monohydrate) may be used as a reference standard in determining the amount of lactose (monohydrate) present in food samples using high performance anion exchange chromatography (HPIEC) with pulsed amperometric detection.[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3265 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of trans-galactooligosaccharides in selected food products by ion-exchange chromatography: collaborative study
Slegte.DJ
Journal of AOAC (Association of Official Analytical Chemists) International, 85, 417-423 (2002)
Fan Zhou et al.
Scientific reports, 1, 78-78 (2012-02-23)
The invisibility cloak has been a long-standing dream for many researchers over the decades. Using transformation optics, a three-dimensional (3D) object is perceived as having a reduced number of dimensions, making it "undetectable" judging from the scattered field12345. Despite successful
Tamás Firkala et al.
Journal of pharmaceutical and biomedical analysis, 107, 318-324 (2015-02-01)
This paper reports the application of surface enhanced Raman chemical imaging (SER-CI) as a potentially non-destructive quantitative analytical method for the investigation of model pharmaceutical formulations containing the active pharmaceutical ingredient (API) in low concentrations (0.5-2%). The application of chemometric
Akhtar Siddiqui et al.
Journal of pharmaceutical sciences, 103(9), 2819-2828 (2014-03-04)
The objective of this study was to develop powder X-ray diffraction (XRPD) chemometric model for quantifying crystalline tacrolimus from solid dispersion (SD). Three SDs (amorphous tacrolimus component) with varying drug to excipient ratios (24.4%, 6.7%, and 4.3% drug) were prepared.
J Nijdam et al.
Colloids and surfaces. B, Biointerfaces, 123, 53-60 (2014-09-30)
Segregation of the protein bovine serum albumin (BSA) and lactose in thin aqueous films during drying was investigated by examining the composition of the dried films using inverse micro Raman spectroscopy (IMRS) and X-ray photoelectron spectroscopy (XPS) sputter-depth profiling. The

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. Rockville, MD
    • Review 1
    • Votes 0
    4 out of 5 stars.

    Which value is certified?

    When I'm looking to purchase certified reference materials to be used in our interlab comparison studies, we need to see which value is certified, but this catalog entry doesn't provide that information. And there is no CoA to evaluate.

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for your review. We encourage customers who experience problems with our products to call or email us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service