Merck
  • Synthesis and quantitative analysis of plasma-targeted metabolites of catechin and epicatechin.
Synthesis and quantitative analysis of plasma-targeted metabolites of catechin and epicatechin.
Journal of agricultural and food chemistry (2015-02-12)
Jack W Blount, Benjamin W Redan, Mario G Ferruzzi, Bradley L Reuhs, Bruce R Cooper, John S Harwood, Vladimir Shulaev, Giulio Pasinetti, Richard A Dixon
ABSTRACT
Grape seed polyphenolic extract (GSPE) rich in the flavan-3-ols (+)-catechin and (-)-epicatechin beneficially modulates Alzheimer's Disease phenotypes in animal models. The parent molecules in the extract are converted to a series of methylated and glucuronidated derivatives. To fully characterize these metabolites and establish a robust quantitative assay of their levels in biological fluids, we have implemented a partial synthetic approach utilizing chemical methylation followed by enzymatic glucuronidation. Liquid chromatography/time-of-flight mass spectrometry (LC-TOF-MS) and nuclear magnetic resonance (NMR) spectroscopy were used to assign unequivocal structures to the compounds. An analytical method using solid-phase extraction and LC-MS/MS in selective reaction monitoring mode (SRM) was validated for their quantitation in plasma. These studies provide a basis for improvements in future work on the bioavailability, metabolism, and mechanism of action of metabolites derived from dietary flavan-3-ols in a range of interventions.
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Epicatechin, primary reference standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%