Merck
Evidence for lymphatics in the developing and adult human choroid.
Investigative ophthalmology & visual science (2015-01-15)
Mark E Koina, Louise Baxter, Samuel J Adamson, Frank Arfuso, Ping Hu, Michele C Madigan, Tailoi Chan-Ling
ABSTRACT
Lymphatics subserve many important functions in the human body including maintenance of fluid homeostasis, immune surveillance, and tumor metastasis. Our aim was to provide structural and phenotypic evidence of lymphatic-like structures in the human choroid, including details of its development. Using multiple-marker immunohistochemistry (IHC), choroids from human fetal eyes (8-26 weeks gestation) and adults (17-74 years) were examined with lymphatic- and vascular-specific markers: prospero homeobox-1 (PROX-1), lymphatic vascular endothelium receptor-1 (LYVE-1), podoplanin, D2-40, endomucin, VEGF-C, vascular endothelial growth factor receptor-3 (VEGFR-3 or Flt4), UEA lectin, platelet endothelial cell adhesion molecule-1 (PECAM-1), CD34, and CD39. Transmission electron microscopy (TEM) was used to establish evidence for choroidal lymphatics, and to provide details of stratification and relative frequency of lymphatics compared to choroidal blood vessels. Immunohistochemistry and TEM indicated a central-to-peripheral topography of lymphatic formation, with numerous blind-ended lymph sacs just external to the choriocapillaris, as well as the presence of infrequent precollector and collector lymphatic channels. Characteristic ultrastructural features of lymphatics in adult human choroid included anchoring filaments, luminal flocculent protein but absence of erythrocytes, fragmented and/or absent basal lamina, absence of intracellular Weibel-Palade bodies, infrequent pericyte ensheathment, and lack of fenestrae. The system of blind-ended initial lymphatic segments seen just external to the fenestrated vessels of the choriocapillaris is ideally placed for recirculating extracellular fluid and strategically placed for immune surveillance. The presence of a system of lymphatic-like channels in the human choroid provides an anatomical basis for antigen presentation in the posterior eye, with a possible route from the eye to the sentinel lymph nodes, similar to that already described for anterior eye lymphatics.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium phosphate, 96%
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Supelco
2-Methylbutane, analytical standard
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
2-Methylbutane, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading