Merck
  • A high boronate avidity monolithic capillary for the selective enrichment of trace glycoproteins.
A high boronate avidity monolithic capillary for the selective enrichment of trace glycoproteins.
Journal of chromatography. A (2015-02-02)
Daojin Li, Yang Li, Xinglin Li, Zijun Bie, Xianghua Pan, Qian Zhang, Zhen Liu
ABSTRACT
Boronate affinity materials, as effective sample enrichment sorbents for glycoproteomic analysis, have attracted increasing attention in recent years. However, most of boronate affinity materials suffer from an apparent limitation, limited binding strength. As a result, extraction of glycoproteins of trace concentration is rather difficult or impossible. In this study, we present a high boronate avidity monolithic capillary. Branched polyethyleneimine (PEI) was used as a scaffold to amplify the number of boronic acid moieties. While 2,4-difluoro-3-formyl-phenylboronic acid (DFFPBA), which exhibited ultrahigh affinity toward cis-diol-containing compounds, was employed as an affinity ligand. Due to the PEI-assisted synergistic multivalent binding, the monolithic column exhibited high boronate avidity toward glycoproteins, with binding constants of 10(-6)-10(-7)M. Such binding strength was the highest among already reported boronic acid-functionalized materials that can be used for glycoproteomic analysis. Besides, the boronate avidity monolithic column exhibited one additional beneficial feature, lowered binding pH (≥6.5). These features greatly favored the selective enrichment of trace glycoproteins from real samples. The feasibility for practical applications was demonstrated with the selective enrichment of trace glycoproteins in human saliva. As compared with other boronate avidity/affinity materials, the boronate avidity monolithic capillary exhibited the best performance.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Glycidyl methacrylate, ≥97.0% (GC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Supelco
Acetic acid, analytical standard
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Glycidyl methacrylate, 97%, contains 100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%