Merck
  • Determination of puerarin in rat plasma using PEGylated magnetic carbon nanotubes by high performance liquid chromatography.
Determination of puerarin in rat plasma using PEGylated magnetic carbon nanotubes by high performance liquid chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-04-29)
Panfeng Yu, Qi Wang, Hongwei Ma, Ji Wu, Shun Shen
ABSTRACT
This paper described a novel application of PEGylated magnetic carbon nanotubes as solid-phase extraction nanosorbents for the determination of puerarin in rat plasma by high performance liquid chromatography (HPLC). A solvothermal method was employed for the synthesis of monodisperse magnetites anchored onto multi-walled carbon nanotubes (MWCNTs@Fe3O4). In order to enhance the water solubility of MWCNTs@Fe3O4 that ensured sufficient contact between nanosorbents and analytes in the sampling procedure, the obtained nanomaterials were further noncovalently functionalized using a phospholipids-polyethylene glycol (DSPE-PEG). The PEGylated MWCNTs@Fe3O4 nanomaterials had an extremely large surface area and exhibit a strong interaction capability for puerarin with π-π stacking interactions. The captured puerarin/nanosorbents were easily isolated from the plasma by placing a magnet, and desorbed by acetonitrile. The experimental variables affecting the extraction efficiency were investigated. The calibration curve of puerarin was linear from 0.01 to 20 μg/ml, and the limit of detection was 0.005 μg/ml. The precisions ranged from 2.7% to 3.5% for within-day measurement, and for between-day variation was in the range of 3.1-5.9%. The method recoveries were acquired from 95.2% to 98.0%. Moreover, the analytical performance obtained by PEGylated magnetic MWCNTs was also compared with that of magnetic MWCNTs. All results showed that our proposed method was an excellent alternative for the analysis of puerarin in rat plasma.
Product Description
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylene glycol, analytical standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol