Merck
  • Full-length transient receptor potential vanilloid 1 channels mediate calcium signals and possibly contribute to osmoreception in vasopressin neurones in the rat supraoptic nucleus.
Full-length transient receptor potential vanilloid 1 channels mediate calcium signals and possibly contribute to osmoreception in vasopressin neurones in the rat supraoptic nucleus.
Cell calcium (2014-12-07)
Taiki Moriya, Rina Shibasaki, Tomohiko Kayano, Nami Takebuchi, Momoko Ichimura, Naoki Kitamura, Atsushi Asano, Yoshinao Z Hosaka, Oksana Forostyak, Alexei Verkhratsky, Govindan Dayanithi, Izumi Shibuya
ABSTRACT
Neurones in the supraoptic nucleus (SON) of the hypothalamus possess intrinsic osmosensing mechanisms, which are lost in transient receptor potential vanilloid 1 (Trpv1)-knock-out mice. The molecular nature of the osmosensory mechanism in SON neurones is believed to be associated with the N-terminal splice variant of Trpv1, although their entire molecular structures have not been hitherto identified. In this study, we sought for TRPV1-related molecules and their function in the rat SON. We performed RT-PCR and immunohistochemistry to detect TRPV1-related molecules in the SON, and patch-clamp and imaging of the cytosolic Ca(2+) concentration ([Ca(2+)]i) to measure responses to osmolality changes and TRPV-related drugs in acutely dissociated SON neurones of rats. RT-PCR analysis revealed full-length Trpv1 and a new N-terminal splice variant, Trpv1_SON (LC008303) in the SON. Positive immunostaining was observed using an antibody against the N-terminal portion of TRPV1 in arginine vasopressin (AVP)-immunoreactive neurones, but not in oxytocin (OT)-immunoreactive neurones. Approximately 20% of SON neurones responded to mannitol (50 mM) with increased action potential firing, inward currents, and [Ca(2+)]i mobilization. Mannitol-induced responses were observed in AVP neurones isolated from AVP-eGFP transgenic rats and identified by GFP fluorescence, but not in OT neurones isolated from OT-mRFP transgenic rats and identified by RFP fluorescence. The mannitol-induced [Ca(2+)]i responses were reversibly blocked by the non-selective TRPV antagonist, ruthenium red (10 μM) and the TRPV1 antagonists, capsazepine (10 μM) and BCTC (10 μM). Although the TRPV1 agonist, capsaicin (100 nM) evoked no response at room temperature, it triggered cationic currents and [Ca(2+)]i elevation when the temperature was increased to 36°C. These results suggest that AVP neurones in the rat SON possess functional full-length TRPV1. Moreover, differences between the responses to capsaicin or hyperosmolality obtained in rat SON neurones and those obtained from dorsal root ganglion neurones or TRPV1-expressing cells indicate that the osmoreceptor expressed in the SON may be a heteromultimer in which TRPV1 is co-assembled with some other, yet unidentified, molecules.
Product Description
USP
Capsaicin, United States Pharmacopeia (USP) Reference Standard
Supelco
Capsaicin, analytical standard
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
BCTC, ≥98% (HPLC)
SAFC
HEPES
Supelco
Octane, analytical standard
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Sigma-Aldrich
Octane, anhydrous, ≥99%
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
[Arg8]-Vasopressin acetate salt, ≥95% (HPLC)
SAFC
HEPES
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ruthenium Red, Technical grade
Sigma-Aldrich
Octane, reagent grade, 98%
Supelco
Capsaicin, Pharmaceutical Secondary Standard; Certified Reference Material
Capsaicin, European Pharmacopoeia (EP) Reference Standard
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%