Merck
  • Characteristics of multi-layer coating formed on commercially pure titanium for biomedical applications.
Characteristics of multi-layer coating formed on commercially pure titanium for biomedical applications.
Materials science & engineering. C, Materials for biological applications (2015-01-13)
Dilek Teker, Faiz Muhaffel, Meryem Menekse, Nevin Gul Karaguler, Murat Baydogan, Huseyin Cimenoglu
ABSTRACT
An innovative multi-layer coating comprising a bioactive compound layer (consisting of hydroxyapatite and calcium titanate) with an underlying titanium oxide layer (in the form of anatase and rutile) has been developed on Grade 4 quality commercially pure titanium via a single step micro-arc oxidation process. Deposition of a multi-layer coating on titanium enhanced the bioactivity, while providing antibacterial characteristics as compared its untreated state. Furthermore, introduction of silver (4.6wt.%) into the multi-layer coating during micro-arc oxidation process imposed superior antibacterial efficiency without sacrificing the bioactivity.
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Calcium acetate monohydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Calcium acetate hydrate, BioUltra, ≥99.0% (calc. on dried material, KT)
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium acetate hydrate, ReagentPlus®, ≥99% (titration), powder
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri