Merck
  • PI3K/mTOR dual inhibitor NVP-BEZ235 decreases Mcl-1 expression and sensitizes ovarian carcinoma cells to Bcl-xL-targeting strategies, provided that Bim expression is induced.
PI3K/mTOR dual inhibitor NVP-BEZ235 decreases Mcl-1 expression and sensitizes ovarian carcinoma cells to Bcl-xL-targeting strategies, provided that Bim expression is induced.
Cancer letters (2014-03-22)
Abdelghani Jebahi, Marie Villedieu, Cécile Pétigny-Lechartier, Emilie Brotin, Marie-Hélène Louis, Edwige Abeilard, Florence Giffard, Marika Guercio, Mélanie Briand, Pascal Gauduchon, Stéphanie Lheureux, Laurent Poulain
ABSTRACT
We previously showed that Bcl-xL and Mcl-1 cooperatively protect platinum-resistant ovarian cancer cells from apoptosis. Here we assessed the anticancer potential of combining ABT-737-induced inhibition of Bcl-xL with Mcl-1 inhibition via PI3K/Akt/mTOR pathway disruption using NVP-BEZ235. NVP-BEZ235 inhibited cell proliferation without inducing apoptosis. It strongly repressed Mcl-1 expression and induced Puma expression in both cell lines tested while differentially modulating Bim between the two. Interestingly, NVP-BEZ235 efficiently sensitized ovarian carcinoma cells to ABT-737, provided that Bim expression was induced. Moreover, inhibiting the ERK1/2 pathway restored Bim expression and sensitized low Bim-expressing cancer cells to the BEZ235/ABT-737 treatment.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Propidium iodide solution