Merck
  • Association between microalbuminuria predicting in-stent restenosis after myocardial infarction and cellular senescence of endothelial progenitor cells.
Association between microalbuminuria predicting in-stent restenosis after myocardial infarction and cellular senescence of endothelial progenitor cells.
PloS one (2015-04-16)
Hisanobu Ota, Naofumi Takehara, Tatsuya Aonuma, Maki Kabara, Motoki Matsuki, Atsushi Yamauchi, Toshiharu Takeuchi, Jun-ichi Kawabe, Naoyuki Hasebe
ABSTRACT
Relationship between microalbuminuria and worse outcome of coronary artery disease patients is discussed, but its underlying pathophysiological mechanism remains unclear. We investigated the role of microalbuminuria to the function of endothelial progenitor cells (EPCs), that might affect to outcome of acute myocardial infarction (AMI) patients. Forty-five AMI patients were divided into two groups according to their urinary albumin excretion: normal (n = 24) and microalbuminuria (>30 mg/day, n = 21). At day-2 and day-7 after AMI onset, circulating-EPCs (CD34+ Flk1+) were quantified by flow cytometry. The number of lectin-acLDL-positive cultured-EPCs immobilized on fibronectin was determined. To assess the cellular senescence of cultured-EPCs, the expression level of sirtuin-1 mRNA and the number of SA-β-gal positive cell were evaluated. Angiographic late in-stent loss after percutaneous coronary intervention (PCI) was evaluated at a six-month follow-up. No significant differences in coronary risk and the extent of myocardial damage were observed between the two groups. Late in-stent loss at the six-month follow-up was significantly higher in the microalbuminuria group (normal:microalbuminuria = 0.76±0.34:1.18±0.57 mm, p=0.021). The number of circulating-EPCs was significantly increased in microalbuminuria group at day-7, however, improved adhesion of EPCs was observed in normal group but not in microalbuminuria group from baseline to day-7 (+3.1±8.3:-1.3±4.4%: p<0.05). On the other hand, in microalbuminuria group at day-7, the level of sirtuin-1 mRNA expression of cultured-EPCs was significantly decreased (7.1±8.9:2.5±3.7 fold, p<0.05), which was based on the negative correlation between the level of sirtuin-1 mRNA expression and the extent of microalbuminuria. The ratio of SA-β-gal-positive cells in microalbuminuria group was increased compared to that of normal group. Microalbuminuria in AMI patients is closely associated with functional disorder of EPCs via cellular senescence, that predicts the aggravation of coronary remodeling after PCI.
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, 97%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vascular Endothelial Growth Factor human, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, BioReagent, suitable for fluorescence, ≥98.0% (TLC)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer