Merck
  • Development of phage immuno-loop-mediated isothermal amplification assays for organophosphorus pesticides in agro-products.
Development of phage immuno-loop-mediated isothermal amplification assays for organophosphorus pesticides in agro-products.
Analytical chemistry (2014-08-20)
Xiude Hua, Wei Yin, Haiyan Shi, Ming Li, Yanru Wang, Hong Wang, Yonghao Ye, Hee Joo Kim, Shirley J Gee, Minghua Wang, Fengquan Liu, Bruce D Hammock
ABSTRACT
Two immuno-loop-mediated isothermal amplification assays (iLAMP) were developed by using a phage-borne peptide that was isolated from a cyclic eight-peptide phage library. One assay was used to screen eight organophosphorus (OP) pesticides with limits of detection (LOD) between 2 and 128 ng mL(-1). The iLAMP consisted of the competitive immuno-reaction coupled to the LAMP reaction for detection. This method provides positive results in the visual color of violet, while a negative response results in a sky blue color; therefore, the iLAMP allows one to rapidly detect analytes in yes or no fashion. We validated the iLAMP by detecting parathion-methyl, parathion, and fenitrothion in Chinese cabbage, apple, and greengrocery, and the detection results were consistent with the enzyme-linked immunosorbent assay (ELISA). In conclusion, the iLAMP is a simple, rapid, sensitive, and economical method for detecting OP pesticide residues in agro-products with no instrumental requirement.
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Paraoxon-ethyl, ≥90%, oil
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Supelco
Paraoxon-ethyl, PESTANAL®, analytical standard
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Supelco
Cyanophos, PESTANAL®, analytical standard
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Supelco
Dimethoate, PESTANAL®, analytical standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M