Merck
  • Longstanding complex regional pain syndrome is associated with activating autoantibodies against alpha-1a adrenoceptors.
Longstanding complex regional pain syndrome is associated with activating autoantibodies against alpha-1a adrenoceptors.
Pain (2014-09-25)
Eric Dubuis, Victoria Thompson, Maria Isabella Leite, Franz Blaes, Christian Maihöfner, David Greensmith, Angela Vincent, Nicolas Shenker, Anoop Kuttikat, Martin Leuwer, Andreas Goebel
ABSTRACT
Complex regional pain syndrome (CRPS) is a limb-confined posttraumatic pain syndrome with sympathetic features. The cause is unknown, but the results of a randomized crossover trial on low-dose intravenous immunoglobulins (IVIG) treatment point to a possible autoimmune mechanism. We tested purified serum immunoglobulin G (IgG) from patients with longstanding CRPS for evidence of antibodies interacting with autonomic receptors on adult primary cardiomyocytes, comparing with control IgG from healthy and diseased controls, and related the results to the clinical response to treatment with low-dose IVIG. We simultaneously recorded both single-cell contractions and intracellular calcium handling in an electrical field. Ten of 18 CRPS preparations and only 1/57 control preparations (P<0.0001) increased the sensitivity of the myocytes to the electric field, and this effect was abrogated by preincubation with α-1a receptor blockers. By contrast, effects on baseline calcium were blocked by preincubation with atropine. Interestingly, serum-IgG preparations from all 4 CRPS patients who had responded to low-dose IVIG with meaningful pain relief were effective in these assays, although 4/8 of the nonresponders were also active. To see if there were antibodies to the α-1a receptor, CRPS-IgG was applied to α-1a receptor-transfected rat-1 fibroblast cells. The CRPS serum IgG induced calcium flux, and fluorescence-activated cell sorting showed that there was serum IgG binding to the cells. The results suggest that patients with longstanding CRPS have serum antibodies to α-1a receptors, and that measurement of these antibodies may be useful in the diagnosis and management of the patients.
Product Description
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material