Merck
  • An outbreak of blaOXA-51-like- and blaOXA-66-positive Acinetobacter baumannii ST208 in the emergency intensive care unit.
An outbreak of blaOXA-51-like- and blaOXA-66-positive Acinetobacter baumannii ST208 in the emergency intensive care unit.
Journal of medical microbiology (2014-08-22)
Satomi Asai, Kazuo Umezawa, Hideo Iwashita, Toshio Ohshima, Maya Ohashi, Mika Sasaki, Hideki Hayashi, Mari Matsui, Keigo Shibayama, Sadaki Inokuchi, Hayato Miyachi
ABSTRACT
A series of clinical isolates of drug-resistant (DR) Acinetobacter baumannii with diverse drug susceptibility was detected from eight patients in the emergency intensive care unit of Tokai University Hospital. The initial isolate was obtained in March 2010 (A. baumannii Tokai strain 1); subsequently, seven isolates were obtained from patients (A. baumannii Tokai strains 2-8) and one isolate was obtained from an air-fluidized bed used by five of the patients during the 3 months from August to November 2011. The isolates were classified into three types of antimicrobial drug resistance patterns (RRR, SRR and SSR) according to their susceptibility (S) or resistance (R) to imipenem, amikacin and ciprofloxacin, respectively. Genotyping of these isolates by multilocus sequence typing revealed one sequence type, ST208, whilst that by a DiversiLab analysis revealed two subtypes. All the isolates were positive for blaOXA-51-like and blaOXA-66, as assessed by PCR and DNA sequencing. A. baumannii Tokai strains 1-8 and 10 (RRR, SRR and SSR) had quinolone resistance-associated mutations in gyrA/parC, as revealed by DNA sequencing. The ISAba1 upstream of blaOXA-51-like and aminoglycoside resistance-associated gene, armA, were detected in A. baumannii Tokai strains 1-7 and 10 (RRR and SRR) as assessed by PCR. Among the genes encoding resistance-nodulation-division family pumps (adeB, adeG and adeJ) and outer-membrane porins (oprD and carO), overexpression of adeB and adeJ and suppression of oprD and carO were seen in isolates of A. baumannii Tokai strain 2 (RRR), as assessed by real-time PCR. Thus, the molecular characterization of a series of isolates of DR A. baumannii revealed the outbreak of ST208 and diverse antimicrobial drug susceptibilities, which almost correlated with differential gene alterations responsible for each type of drug resistance.
Product Description
Supelco
Isopropyl myristate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tobramycin, Pharmaceutical Secondary Standard; Certified Reference Material
Ceftazidime, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Isopropyl myristate, ≥90% (GC)
Sigma-Aldrich
Tobramycin, Aminoglycoside antibiotic
Supelco
Aztreonam, analytical standard
Sigma-Aldrich
Isopropyl myristate, ≥98%
Sigma-Aldrich
Isopropyl myristate, 98%
USP
Tobramycin, United States Pharmacopeia (USP) Reference Standard
Tobramycin, European Pharmacopoeia (EP) Reference Standard
USP
Isopropyl myristate, United States Pharmacopeia (USP) Reference Standard
USP
Meropenem, United States Pharmacopeia (USP) Reference Standard