Merck
  • Improvement of the activation of lipase from Candida rugosa following physical and chemical immobilization on modified mesoporous silica.
Improvement of the activation of lipase from Candida rugosa following physical and chemical immobilization on modified mesoporous silica.
Materials science & engineering. C, Materials for biological applications (2014-12-11)
Chunfeng Wang, Yanjing Li, Guowei Zhou, Xiaojie Jiang, Yunqiang Xu, Zhaosheng Bu
ABSTRACT
Lipase from Candida rugosa (CRL) was chemically and physically immobilized onto four types of rod-shaped mesoporous silica (RSMS). RSMS prepared using surfactant P123 and poly(ethylene glycol) as co-templates was functionalized with (3-aminopropyl)triethoxysilane (APTES) to obtain P-RSMS by post-synthesis grafting. Tetraethoxysilane was hydrothermally co-condensed with APTES to obtain C-RSMS. A two-step process using APTES and glutaraldehyde was also performed to obtain G-RSMS. The effects of modification methods (including post-synthesis grafting and co-condensation) and glutaraldehyde on the mesoscopic order, interplanar spacing d100, cell parameter a0, mesoporous structure, and wall thickness of RSMS were studied in detail. Results showed that all samples were mesoporous materials with 2D mesostructures (p6mm). Pore size and d100 decreased, whereas the wall thickness increased after different modifications. CRL was used as a model enzyme to determine the effect of physical and chemical adsorption on loading amount and enzymatic activity. The possible mechanism of CRL immobilization on G-RSMS by chemical adsorption was systematically investigated. The chemical immobilization of CRL on G-RSMS increased the loading amount, hydrolytic activity, thermal stability, and reusability. Moreover, immobilized CRL was employed to catalyze the resolution of 2-octanol by esterification with caprylic acid. The enantiomeric excess of 2-octanol was 45.8% when the reaction was catalyzed by G-RSMS-CRL and decreased to about 38%-39% using the physically immobilized CRL, after 48 h of reaction in hexane.
Product Description
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triacetin, 99%, FCC, FG
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Triacetin, 99%
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Supelco
Triacetin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Supelco
Triacetin, analytical standard
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%