Merck
  • L-DOPA-induced dyskinesia in a rat model of Parkinson's disease is associated with the fluctuational release of norepinephrine in the sensorimotor striatum.
L-DOPA-induced dyskinesia in a rat model of Parkinson's disease is associated with the fluctuational release of norepinephrine in the sensorimotor striatum.
Journal of neuroscience research (2014-07-01)
Yong Wang, Hui Sheng Wang, Tao Wang, Chen Huang, Jian Liu
ABSTRACT
L-3,4-dihydroxyphenylalanine (L-DOPA)-induced dyskinesia (LID) is the most common complication of standard L-DOPA therapy for Parkinson's disease experienced by most parkinsonian patients. LID is associated with disruption of dopaminergic homeostasis in basal ganglia following L-DOPA administration. Norepinephrine (NE) is another important catecholaminergic neurotransmitter that is also believed to be involved in the pathogenesis of LID. This study compared NE release in the ipsilateral sensorimotor striatum of dyskinetic and nondyskinetic 6-hydroxydopamine-lesioned hemiparkinsonian rats treated chronically with L-DOPA. After L-DOPA injection, the time-course curves of NE levels in the sensorimotor striatum were significantly different between dyskinetic and nondyskinetic rats. Several metabolic kinetic parameters of NE levels were also differentially expressed between the two groups. In comparison with nondyskinetic rats, the ΔCmax of NE was significantly higher in dyskinetic rats, whereas Tmax and t1/2 of NE were significantly shorter. Intrastriatal perfusion of NE into the lesioned sensorimotor striatum revealed a moderate dyskinesia in dyskinetic rats, which was similar to the dyskinetic behavior after L-DOPA administration. The L-DOPA-related dyskinetic behavior was inhibited significantly by a further pretreatment of noradrenergic neurotoxin N-​(2-​chloroethyl)​-​N-​ethyl-​2-​bromobenzylamine or intrastriatal administration of the α2 -adrenoceptor antagonist idazoxan, accompanied by significant changes in metabolic kinetic parameters of NE in the sensorimotor striatum. The results provide evidence to support the correlation between abnormal NE neurotransmission and the induction of LID and suggest that the aberrant change of the quantitative and temporal releasing of NE in the sensorimotor striatum might play an important role in the pathogenesis of LID.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Levodopa, United States Pharmacopeia (USP) Reference Standard
SAFC
Sodium chloride solution, 5 M
Levodopa, European Pharmacopoeia (EP) Reference Standard
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benserazide hydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, solid
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis