Merck
  • Both leukaemic and normal peripheral B lymphoid cells are highly sensitive to the selective pharmacological inhibition of prosurvival Bcl-2 with ABT-199.
Both leukaemic and normal peripheral B lymphoid cells are highly sensitive to the selective pharmacological inhibition of prosurvival Bcl-2 with ABT-199.
Leukemia (2014-01-10)
S L Khaw, D Mérino, M A Anderson, S P Glaser, P Bouillet, A W Roberts, D C S Huang
ABSTRACT
Overexpression of the prosurvival protein Bcl-2 marks many B-lymphoid malignancies and contributes to resistance to many commonly used chemotherapeutic agents. The first effective BH3 mimetic inhibitors of Bcl-2, ABT-737 and navitoclax, also target Bcl-xL, causing dose-limiting thrombocytopenia. This prompted the development of the Bcl-2-selective antagonist, ABT-199. Here we show that in lymphoid cells, ABT-199 specifically causes Bax/Bak-mediated apoptosis that is triggered principally by the initiator BH3-only protein Bim. As expected, malignant cells isolated from patients with chronic lymphocytic leukaemia are highly sensitive to ABT-199. However, we found that normal, untransformed mature B cells are also highly sensitive to ABT-199, both in vitro and in vivo. By contrast, the B-cell precursors are largely spared, as are cells of myeloid origin. These results pinpoint the probable impact of the pharmacological inhibition of Bcl-2 by ABT-199 on the normal mature haemopoietic cell lineages in patients, and have implications for monitoring during ABT-199 therapy as well as for the clinical utility of this very promising targeted agent.
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
4-Hydroxytamoxifen, (E) and (Z) isomers (50:50), analytical standard
Supelco
4-Hydroxytamoxifen, analytical standard, (E) and (Z) isomers (50:50)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
SAFC
BIS-TRIS
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS