Merck
  • Automated analytical standard production with supercritical fluid chromatography for the quantification of bioactive C17-polyacetylenes: a case study on food processing waste.
Automated analytical standard production with supercritical fluid chromatography for the quantification of bioactive C17-polyacetylenes: a case study on food processing waste.
Food chemistry (2014-07-21)
Sebastiaan Bijttebier, Els D'Hondt, Bart Noten, Nina Hermans, Sandra Apers, Vassiliki Exarchou, Stefan Voorspoels
ABSTRACT
Food processing enterprises produce enormous amounts of organic waste that contains valuable phytochemicals (e.g. C17-polyacetylenes). Knowledge on the phytochemicals content is a first step towards valorisation. Quantification of C17-polyacetylenes is however often hampered by the lack of commercially available standards or by tedious multistep in-house standard production procedures. In the current study, a new and straightforward supercritical fluid chromatography purification procedure is described for the simultaneous production of 2 analytical C17-polyacetylene standards. Respectively, 5 and 6 mg of falcarinol and falcarindiol were purified in 17 h on analytical scale. After confirming the identity and quality (97% purity) by Nuclear Magnetic Resonance, accurate mass-Mass Spectrometry (am-MS) and Photo Diode Array (PDA) detection the C17-polyacetylene standards were used for the analysis of industrial vegetable waste with Liquid Chromatography coupled to PDA and am-MS detection. Measurements showed varying concentrations of C17-polyacetylenes in the organic waste depending on its nature and origin.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis