Merck
  • Flavonoid-based pH-responsive hydrogels as carrier of unstable drugs in oxidative conditions.
Flavonoid-based pH-responsive hydrogels as carrier of unstable drugs in oxidative conditions.
Pharmaceutical development and technology (2013-12-03)
Umile Gianfranco Spizzirri, Giuseppe Cirillo, Manuela Curcio, Nevio Picci, Francesca Iemma
ABSTRACT
In this study, pH-responsive hydrogels, synthesized by the coupling reaction of polyacrylic acid and catechin, are proposed as carriers of oxidable drugs toward the GI tract. The presence of polyphenolic moieties in the network gives the polymers properties suitable for the release of unstable drugs in oxidative conditions. The characterization of the hydrogels is obtained by means of morphological and physico-chemical analyses, antioxidant assays and evaluation of the swelling behavior in media simulating the gastric (pH 1.0) and the intestinal (pH 7.4) tracts. The hydrogels are tested as pH-responsive carriers in in vitro release studies of folic acid and thiamine, two model drugs easily degraded by oxidative conditions simulated by UV irradiation and t-butyl hydroperoxide treatment, respectively. Results show that catechin-based carriers are able to control the release of drugs at different pH values, giving a remarkable improvement in the stability of the therapeutics.
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Hexylene glycol, puriss., ≥99.0% (GC)
Sigma-Aldrich
Hexylene glycol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Hexylene glycol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
SAFC
Hexylene glycol
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Hexylene glycol, 99%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
1,6-Hexanediol, 97%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium persulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
1,6-Hexanediol, 99%
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material