Merck
  • Evaluation of quadrupole-time-of-flight mass spectrometry in comprehensive two-dimensional gas chromatography.
Evaluation of quadrupole-time-of-flight mass spectrometry in comprehensive two-dimensional gas chromatography.
Journal of chromatography. A (2014-09-03)
Blagoj Mitrevski, Philip J Marriott
ABSTRACT
Quadrupole time-of-flight mass spectrometry (Q-TOFMS) has been evaluated with respect to its applicability in comprehensive two-dimensional gas chromatography (GC×GC). At a maximum acquisition frequency, while approximately 50 full accurate mass spectra on disk were acquired per s (50Hz) in scan mode, the sampling rate in target mode (MS/MS) was strongly dependent on the number of target ions selected. The number of selected precursor ions per time window proportionally decreased the acquisition rate for each ion; one precursor ion ≅31.35Hz; two ions ≅16.68Hz; and for 8 precursor ions, a sampling rate of just 4.18Hz was found. When Q-TOFMS was used in simultaneous mode, where in addition to the acquisition of target ion MS/MS signals, it also collects the full mass spectrum, sampling rates were even lower. It is demonstrated that Q-TOFMS generates sufficient data points over each peak in GC×GC operation in scan mode using TOFMS acquisition only, or is able to collect sufficient data points for relatively wide chromatographic peaks (≥600ms) in the target mode (MS/MS), however only if one or two precursor ions are selected per time window. Mass accuracy was found to perform within specification (<5ppm), even for the fastest acquisition operation (50Hz). Spectral deconvolution is demonstrated to work better in GC×GC than in 1D GC mode. Data visualisation in target GC×GC mode presents difficulties when there are overlapping target windows comprising different numbers of precursor ions.
Sigma-Aldrich
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D)
Supelco
Tributyl phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Triphenyl phosphate solution, NMR reference standard, 3 mM in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Triphenyl phosphate, analytical standard
Sigma-Aldrich
Tributyl phosphate, 97%
Supelco
Tributyl phosphate, for extraction analysis, ≥99.0% (GC)
Sigma-Aldrich
Triphenyl phosphate, ≥99%
Sigma-Aldrich
2-Methylheptane, 98%
Sigma-Aldrich
Tributyl phosphate, ≥99%
Tri-n-butyl phosphate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tributyl phosphate, puriss., ≥99.0% (GC)
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Dicyclohexylamine, 99%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Acetone, analytical standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%