Skip to Content
Merck
All Photos(1)

Key Documents

708968

Sigma-Aldrich

Triphenyl phosphate solution

NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D)

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$952.00
1 PKG
$952.00

$952.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 PKG
$952.00
1 PKG
$952.00

About This Item

Linear Formula:
(C6H5O)3PO
Molecular Weight:
326.28
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.12

$952.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

grade

NMR reference standard

form

solution

concentration

0.0485 M in acetone-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 7 in.
5 mm × 8 in.

suitability

suitable for NMR (reference standard)

SMILES string

O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3

InChI

1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H

InChI key

XZZNDPSIHUTMOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

31P sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davide Savy et al.
Molecules (Basel, Switzerland), 20(11), 19958-19970 (2015-11-12)
The molecular composition of water-soluble lignins isolated from four non-food bioenergy crops (cardoon CAR, eucalyptus EUC, and two black poplars RIP and LIM) was characterized in detail, and their potential bioactivity towards maize germination and early growth evaluated. Lignins were
Sin-Ying Koou et al.
Parasites & vectors, 7, 471-471 (2014-10-11)
In Singapore, dose-response bioassays of Aedes aegypti (L.) adults have been conducted, but the mechanisms underlying resistance to insecticides remain unclear. In this study, we evaluated insecticide resistance and its underlying mechanism in field populations of Ae. aegypti adults. Seven
M I Cervera et al.
Analytical and bioanalytical chemistry, 406(27), 6843-6855 (2014-05-16)
An atmospheric pressure chemical ionization source has been used to enhance the potential of gas chromatography coupled with quadrupole time-of-flight (QTOF) mass spectrometry (MS) for screening and quantification purposes in pesticide residue analysis. A screening method developed in our laboratory
Francesc A Esteve-Turrillas et al.
PloS one, 10(7), e0134042-e0134042 (2015-07-28)
Bioconjugate preparation is a fundamental step for antibody generation and immunoassay development to small chemical compounds. For analytical targets holding in their structure an aryl halogen atom, cross-coupling reactions may be a simple and efficient way to obtain functionalized derivatives;
Boris V Krivoshiev et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(8), 2124-2132 (2015-09-08)
Flame retardants are ubiquitously used chemicals that have been shown to contaminate environments. Toxicological data is largely limited, with little insight into their molecular modes of action that may give rise to their toxic phenotypes. Such insight would aid more

Questions

1–2 of 2 Questions  
  1. What is the volume for the 8 inch length tube of product 708968, Triphenyl phosphate solution, NMR reference standard, 0.0485M in acetone-d6 (99.9 atom % D)

    1 answer
    1. The volume in the 8 inch length tube of product 708968, Triphenyl phosphate solution, NMR reference standard, 0.0485M in acetone-d6 (99.9 atom % D), is 0.7 ml.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service