Merck
  • Global mass spectrometry and transcriptomics array based drug profiling provides novel insight into glucosamine induced endoplasmic reticulum stress.
Global mass spectrometry and transcriptomics array based drug profiling provides novel insight into glucosamine induced endoplasmic reticulum stress.
Molecular & cellular proteomics : MCP (2014-08-17)
Ana Sofia Carvalho, Helena Ribeiro, Paula Voabil, Deborah Penque, Ole N Jensen, Henrik Molina, Rune Matthiesen
ABSTRACT
We investigated the molecular effects of glucosamine supplements, a popular and safe alternative to nonsteroidal anti-inflammatory drugs, for decreasing pain, inflammation, and maintaining healthy joints. Numerous studies have reported an array of molecular effects after glucosamine treatment. We questioned whether the differences in the effects observed in previous studies were associated with the focus on a specific subproteome or with the use of specific cell lines or tissues. To address this question, global mass spectrometry- and transcription array-based glucosamine drug profiling was performed on malignant cell lines from different stages of lymphocyte development. We combined global label-free MS-based protein quantitation with an open search for modifications to obtain the best possible proteome coverage. Our data were largely consistent with previous studies in a variety of cellular models. We mainly observed glucosamine induced O-GlcNAcylation/O-GalNAcylation (O-HexNAcylation); however, we also observed global and local changes in acetylation, methylation, and phosphorylation. For example, our data provides two additional examples of "yin-yang" between phosphorylation and O-HexNAcylation. Furthermore, we mapped novel O-HexNAc sites on GLU2B and calnexin. GLU2B and calnexin are known to be located in the endoplasmic reticulum (ER) and involved in protein folding and quality control. The O-HexNAc sites were regulated by glucosamine treatment and correlated with the up-regulation of the ER stress marker GRP78. The occupancy of O-HexNAc on GLU2B and calnexin sites differed between the cytosolic and nuclear fractions with a higher occupancy in the cytosolic fraction. Based on our data we propose the hypothesis that O-HexNAc either inactivates calnexin and/or targets it to the cytosolic fraction. Further, we hypothesize that O-HexNAcylation induced by glucosamine treatment enhances protein trafficking.
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)