Skip to Content
Merck
All Photos(6)

Key Documents

M2069

Sigma-Aldrich

D-(+)-Mannose

≥99% (GC), wood

Synonym(s):

D-Mannopyranose

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$69.70
100 G
$247.00
500 G
$854.00

$69.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$69.70
100 G
$247.00
500 G
$854.00

About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1564373
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

$69.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

wood

Quality Level

Assay

≥99% (GC)

form

powder

technique(s)

gas chromatography (GC): suitable

color

white to off-white

mp

133-140 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

room temp

SMILES string

OC[C@H]1OC(O)[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5+,6?/m1/s1

InChI key

WQZGKKKJIJFFOK-QTVWNMPRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mannose is a monosaccharide.

Application

D-(+)-Mannose, a C-2 epimer of D-glucose, is used in the formation of glycan structure and glycosylation.
D-Mannose has been used in a study to assess the synthesis of a family of amphiphilic glycopolymers. [1] It has also been used in a study to investigate the early detection of bronchiolitis obliterans after lung transplantation. [2]

Biochem/physiol Actions

Mannose, a six-carbon carbohydrate, is the C-2 epimer of glucose and a critical sugar for protein glycosylation. Mannose can also be utilized by the brain as an alternative energy source.

Other Notes

From spruce, birch, or beech wood
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Quality

Predominantly α-isomer

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabian Suriano et al.
Biomaterials, 31(9), 2637-2645 (2010-01-16)
Polymers bearing pendant carbohydrates have a variety of biomedical applications especially in the area of targeted drug delivery. Here we report the synthesis of a family of amphiphilic block glycopolymers containing d glucose, d galactose and d mannose via metal-free
Bithi Chatterjee et al.
Blood, 120(10), 2011-2020 (2012-07-14)
Dendritic cells (DCs) can capture extracellular antigens and load resultant peptides on to MHC class I molecules, a process termed cross presentation. The mechanisms of cross presentation remain incompletely understood, particularly in primary human DCs. One unknown is the extent
Steven J Budd et al.
Respiratory research, 13, 56-56 (2012-07-06)
Long-term lung allograft survival is limited by bronchiolitis obliterans syndrome (BOS). Mannose binding lectin (MBL) belongs to the innate immune system, participates in complement activation, and may predispose to graft rejection. We investigated mannose binding (MBL) during cold ischemia and
Troels R Kjaer et al.
Infection and immunity, 81(2), 452-459 (2012-11-28)
The three human ficolins (H-, L-, and M-ficolins) and mannan-binding lectin are pattern recognition molecules of the innate immune system mediating activation of the lectin pathway of the complement system. These four human proteins bind to some microorganisms and may
Conghui Liu et al.
Frontiers in immunology, 11, 1379-1379 (2020-08-15)
Cluster of differentiation 63 (CD63), a four-transmembrane glycoprotein in the subfamily of tetraspanin, has been widely recognized as a gateway from the infection of foreign invaders to the immune defense of hosts. Its role in Pacific oyster Crassostrea gigas is

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service