Merck
  • A novel surface-confined glucaminium-based ionic liquid stationary phase for hydrophilic interaction/anion-exchange mixed-mode chromatography.
A novel surface-confined glucaminium-based ionic liquid stationary phase for hydrophilic interaction/anion-exchange mixed-mode chromatography.
Journal of chromatography. A (2014-08-19)
Lizhen Qiao, Shuangyuan Wang, Hua Li, Yuanhong Shan, Abo Dou, Xianzhe Shi, Guowang Xu
ABSTRACT
Glucaminium-based ionic liquids are a new class of recently developed ionic liquids and prepared by functionalizing the amine group of N-methyl-d-glucamine, which renders them good hydrophilicity due to the presence of the glucose structure and charged quaternary ammonium group. In the present study, a glucaminium-based ionic liquid N,N-diallyl-N-methyl-d-glucaminium bromide was synthesized and subsequently bonded to the surface of 3-mercaptopropyl modified silica materials through "thiol-ene" click chemistry. The obtained stationary phase was characterized by elemental analysis and infrared spectroscopy, and then packed as a HPLC column. A mixture of five nucleosides was used to characterize the separation performance of the obtained column under HILIC mode and the column efficiency was determined with cytidine as the test solute, reaching 80,000plates/m. Then, the retention behavior was evaluated by investigating the effect of various chromatographic factors on retention of different types of solutes, and the results revealed that the developed surface-confined glucaminium-based ionic liquid stationary phase exhibited a hydrophilic interaction/anion-exchange mixed-mode retention mechanism. Finally, two mixtures of nucleotides and flavonoids were separated on the glucaminium-based ionic liquid column, respectively under hydrophilic interaction and hydrophilic interaction/anion-exchange mixed-mode chromatography. In conclusion, the multimodal retention capabilities of the glucaminium-based ionic liquid column could offer a wider range of retention behavior and flexible selectivity toward polar and hydrophilic compounds.
Product Description
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Adenosine 5′-diphosphate disodium salt, ≥90%
Sigma-Aldrich
Cytidine, 99%
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Cytidine 5′-monophosphate, Sigma Grade, ≥99% (HPLC), synthetic, powder
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Sigma-Aldrich
Meglumine, 99.0-100.5% dry basis, meets USP testing specifications
Sigma-Aldrich
Adenosine 5′-monophosphate monohydrate, from yeast, ≥97%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Guanosine 5′-monophosphate disodium salt hydrate, from yeast, ≥99%
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Supelco
Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Inosine 5′-diphosphate sodium salt, ≥96%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Ammonium formate, reagent grade, 97%