Merck
  • Impaired gamma delta T cell-derived IL-17A and inflammasome activation during early respiratory syncytial virus infection in infants.
Impaired gamma delta T cell-derived IL-17A and inflammasome activation during early respiratory syncytial virus infection in infants.
Immunology and cell biology (2014-10-01)
Huaqiong Huang, Jordy Saravia, Dahui You, Aaron J Shaw, Stephania A Cormier
ABSTRACT
Respiratory syncytial virus (RSV) infection remains a significant global health burden disproportionately affecting infants and leading to long-term lung disease. Interleukin (IL)-17A has been shown to be involved in regulating viral and allergic lung inflammatory responses, which has led to a more recent interest in its role in RSV infection. Using a neonatal mouse model of RSV, we demonstrate that neonates fail to develop IL-17A responses compared with adult mice; the main immediate IL-17A contributor in adults were γδ T cells. Antibody neutralization of IL-17A in adult mice caused increased lung inflammation and airway mucus from RSV, whereas exogenous IL-17A administration to RSV-infected neonates caused decreased inflammation but no change in airway mucus. We also observed a lack of pro-inflammatory cytokine production (IL-1β, IL-6) from infected neonates. Using human cord blood mononuclear cells (CBMCs) and adult peripheral blood mononuclear cells (PBMCs), we compared inflammasome activation by direct retinoic acid-inducible gene I agonism; CBMCs failed to induce pro-inflammatory cytokines or IL-17A(+) γδ T cells compared with PBMCs. Our results indicate that RSV disease severity is in part mediated by a lack of inflammasome activation and IL-17A production in neonates.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)