Merck
  • The molecular mechanism of sporocyteless/nozzle in controlling Arabidopsis ovule development.
The molecular mechanism of sporocyteless/nozzle in controlling Arabidopsis ovule development.
Cell research (2014-11-08)
Baoye Wei, Jinzhe Zhang, Changxu Pang, Hao Yu, Dongshu Guo, Hao Jiang, Mingxin Ding, Zhuoyao Chen, Qing Tao, Hongya Gu, Li-Jia Qu, Genji Qin
ABSTRACT
Ovules are essential for plant reproduction and develop into seeds after fertilization. Sporocyteless/nozzle (SPL/NZZ) has been known for more than 15 years as an essential factor for ovule development in Arabidopsis, but the biochemical nature of SPL function has remained unsolved. Here, we demonstrate that SPL functions as an adaptor-like transcriptional repressor. We show that SPL recruits topless/topless-related (TPL/TPR) co-repressors to inhibit the Cincinnata (CIN)-like Teosinte branched1/cycloidea/PCF (TCP) transcription factors. We reveal that SPL uses its EAR motif at the C-terminal end to recruit TPL/TPRs and its N-terminal part to bind and inhibit the TCPs. We demonstrate that either disruption of TPL/TPRs or overexpression of TCPs partially phenocopies the defects of megasporogenesis in spl. Moreover, disruption of TCPs causes phenotypes that resemble spl-D gain-of-function mutants. These results define the action mechanism for SPL, which along with TPL/TPRs controls ovule development by repressing the activities of key transcription factors. Our findings suggest that a similar gene repression strategy is employed by both plants and fungi to control sporogenesis.
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzyl benzoate, analytical standard
Supelco
Benzyl alcohol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzyl benzoate, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl benzoate, tested according to Ph. Eur.
Sigma-Aldrich
Benzyl benzoate, natural, ≥99%, FG
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl benzoate, meets USP testing specifications
Supelco
Benzyl benzoate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Benzyl benzoate, United States Pharmacopeia (USP) Reference Standard
USP
Benzyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Benzyl benzoate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%