Merck
Evaluation of enantiomeric purity of magnesium-L-aspartate dihydrate.
Journal of pharmaceutical and biomedical analysis (2014-09-30)
Oliver Wahl, Ulrike Holzgrabe
ABSTRACT
Magnesium supplementation in form of organic magnesium salts is a very popular practice. We examined the enantiomeric purity of "Magnesium aspartate dihydrate" monographed in the European Pharmacopeia. A chiral capillary zone electrophoresis using (2-hydroxypropyl)-β-cyclodextrin coupled to laser induced fluorescence detection and a HPLC-fluorescence method with chiral derivatization using o-phthaldialdehyde and N-acetyl-L-cysteine as an orthogonal method were developed and validated. Two batch samples of this substance and three drug products containing the salt were analyzed by means of both methods. The concentration of the D-enantiomer of aspartic acid ranged from 0.03 to 0.12%. Simulations of the synthesis revealed that the d-aspartic acid content is elevated if the dissolution of L-aspartic acid was performed at acidic pH values.
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioXtra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Potassium cyanide, BioUltra, ≥98.0% (AT)
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioXtra, ≥98.0% (T)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology