Merck
  • In vivo evaluation of porous silicon and porous silicon solid lipid nanocomposites for passive targeting and imaging.
In vivo evaluation of porous silicon and porous silicon solid lipid nanocomposites for passive targeting and imaging.
Molecular pharmaceutics (2014-07-01)
Annukka M Kallinen, Mirkka P Sarparanta, Dongfei Liu, Ermei M Mäkilä, Jarno J Salonen, Jouni T Hirvonen, Hélder A Santos, Anu J Airaksinen
ABSTRACT
The use of nanoparticle carriers for the sustained release of cytotoxic drugs in cancer therapy can result in fewer adverse effects and can thus be of great benefit for the patient. Recently, a novel nanocomposite, prepared by the encapsulation of THCPSi nanoparticles within solid lipids (SLN), was developed and characterized as a promising drug delivery carrier in vitro. The present study describes the in vivo evaluation of unmodified THCPSi nanoparticles and THCPSi-solid lipid nanocomposites (THCPSi-SLNCs) as potential drug delivery carriers for cancer therapy by using (18)F radiolabeling for the detection of the particle biodistribution in mice. Passive tumor targeting of (18)F-THCPSis and (18)F-THCPSi-SLNCs by the enhanced permeation and retention effect was investigated in a murine breast cancer model. Encapsulation of THCPSi nanoparticles with solid lipids improved their accumulation in tumors at a 7 week time point (tumor-to-liver ratio 0.10 ± 0.08 and 0.24 ± 0.09% for (18)F-THCPSis and (18)F-THCPSi-SLNCs, respectively).
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
1-Stearoyl-rac-glycerol, ≥99%
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Sodium iodide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium iodide, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Boron, crystalline, 1 cm, 99.7% trace metals basis
Sigma-Aldrich
Boron, crystalline, −60 mesh, 99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
AIDA, solid
Sigma-Aldrich
Boron, ≥95% (boron), amorphous powder
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Boron, monofilament, 50m, diameter 0.1mm
Boron, monofilament, 100m, diameter 0.1mm